Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282795.1 | NP_001269724.1 | 5 | 1781 | |
NM_001282797.1 | NP_001269726.1 | 4 | 1621 | ||
NM_003416.3 | NP_003407.1 | 5 | 1748 | ||
XM_006716654.2 | XP_006716717.1 | 3 | 1512 | ||
XM_006716656.3 | XP_006716719.1 | 3 | 1501 | ||
XM_011517291.2 | XP_011515593.1 | 5 | 1795 | ||
XM_011517292.2 | XP_011515594.1 | 5 | 2129 | ||
XM_011517293.2 | XP_011515595.1 | 5 | 2128 | ||
XM_011517294.2 | XP_011515596.1 | 5 | 1907 | ||
XM_011517295.2 | XP_011515597.1 | 5 | 1688 | ||
XM_011517296.2 | XP_011515598.1 | 5 | 1699 | ||
XM_011517297.2 | XP_011515599.1 | 3 | 1548 | ||
XM_017013813.1 | XP_016869302.1 | 5 | 1792 | ||
XM_017013814.1 | XP_016869303.1 | 4 | 2094 | ||
XM_017013815.1 | XP_016869304.1 | 4 | 2091 | ||
GenBank mRNA | AB209619.1 | - | 2 | 5137 | |
AK300001.1 | - | 5 | 1716 | ||
AK301561.1 | - | 4 | 1582 | ||
AK313745.1 | - | 5 | 1688 | ||
BC058923.1 | - | 5 | 1692 | 4516166 | |
BX647998.1 | - | 5 | 1707 | ||
M29580.1 | - | 5 | 1849 | ||
U71599.1 | - | 1 | 394 |