Gene Transcripts

Gene Symbol
GPR37L1
Entrez Gene ID
9283
Gene Name
G protein-coupled receptor 37 like 1
Gene Aliases
ET(B)R-LP-2, ETBR-LP-2
Chromosome Location
Chr. 1: 202118669 - 202133588 on Build GRCh38
UniGene ID
Hs.132049
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004767.3 NP_004758.3 1 618
GenBank mRNA AK222639.1 - 1 600
AK295576.1 - 3 601
AK313044.1 - 1 645
BC050334.1 - 1 618 5174449
Y16280.1 - 1 606

View Full Product Details