Gene Transcripts

Gene Symbol
ADGRL1
Entrez Gene ID
22859
Gene Name
adhesion G protein-coupled receptor L1
Gene Aliases
CIRL1, CL1, LEC2, LPHN1
Chromosome Location
Chr. 19: 14147737 - 14206245 on Build GRCh38
UniGene ID
Hs.94229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008701.2 NP_001008701.1 17, 18 3341
NM_014921.4 NP_055736.2 16, 17 3326
XM_005259818.3 XP_005259875.1 16, 17 3382
XM_011527796.2 XP_011526098.1 17, 18 3418
XM_011527798.2 XP_011526100.1 16, 17 3403
XM_011527801.2 XP_011526103.1 15, 16 2858
XM_017026475.1 XP_016881964.1 17, 18 3421
XM_017026476.1 XP_016881965.1 18, 19 3531
XM_017026477.1 XP_016881966.1 17, 18 3400
XM_017026478.1 XP_016881967.1 17, 18 3421
XM_017026479.1 XP_016881968.1 17, 18 3421
XM_017026480.1 XP_016881969.1 14, 15 2836
GenBank mRNA AB020628.1 - 17, 18 3343
AF307079.1 - 16, 17 3310
BC007587.1 - 7, 8 1141

View Full Product Details