Gene Transcripts
- Gene Symbol
- SHANK2
- Entrez Gene ID
- 22941
- Gene Name
- SH3 and multiple ankyrin repeat domains 2
- Gene Aliases
- AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3
- Chromosome Location
- Chr. 11: 70467856 - 71252724 on Build GRCh38
- UniGene ID
- Hs.268726
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012309.4 |
NP_036441.2
|
15, 16 |
2051 |
|
NM_133266.4 |
NP_573573.2
|
1, 2 |
248 |
|
NR_110766.1 |
|
1, 2 |
248 |
|
XM_005277932.3 |
XP_005277989.1
|
6, 7 |
1056 |
|
XM_017017387.1 |
XP_016872876.1
|
16, 17 |
2156 |
|
XM_017017388.1 |
XP_016872877.1
|
16, 17 |
2178 |
|
XM_017017389.1 |
XP_016872878.1
|
16, 17 |
2351 |
|
XM_017017390.1 |
XP_016872879.1
|
3, 4 |
3289 |
|
GenBank mRNA |
AB208025.1 |
- |
15, 16 |
2017 |
|
AB208026.1 |
- |
6, 7 |
839 |
|
AB208027.1 |
- |
1, 2 |
226 |
|
AF141901.1 |
- |
1, 2 |
140 |
|
BC093885.1 |
- |
1, 2 |
248 |
|
BC112097.1 |
- |
1, 2 |
248 |
|
BC113876.1 |
- |
1, 2 |
171 |
|
BC114484.1 |
- |
1, 2 |
170 |
|
BC143389.1 |
- |
1, 2 |
248 |
|
DQ152234.1 |
- |
14, 15 |
1939 |
|
View Full Product Details