Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015143.2 | NP_055958.2 | 11 | 1158 | |
XM_011531777.2 | XP_011530079.1 | 11 | 1155 | ||
XM_011531778.1 | XP_011530080.1 | 11 | 951 | ||
XM_011531779.1 | XP_011530081.1 | 10 | 1026 | ||
XM_017007914.1 | XP_016863403.1 | 10 | 1014 | ||
XM_017007915.1 | XP_016863404.1 | 12 | 1172 | ||
XM_017007916.1 | XP_016863405.1 | 11 | 1171 | ||
XM_017007917.1 | XP_016863406.1 | 11 | 947 | ||
GenBank mRNA | AK304239.1 | - | 11 | 1074 | |
AK309465.1 | - | 3 | 231 | ||
BC030054.1 | - | 7 | 716 | 3958447 | |
CR936632.1 | - | 13 | 4736 | ||
D42084.1 | - | 11 | 1066 |