Gene Transcripts

Gene Symbol
RAB39A
Entrez Gene ID
54734
Gene Name
RAB39A, member RAS oncogene family
Gene Aliases
RAB39
Chromosome Location
Chr. 11: 107928475 - 107963482 on Build GRCh38
UniGene ID
Hs.354906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017516.2 NP_059986.1 2 479
GenBank mRNA AK292969.1 - 2 451
BC028064.1 - 2 403 5197104
HY107079.1 - 2 480
X99962.1 - 2 385

View Full Product Details