Gene Transcripts

Gene Symbol
UGGT2
Entrez Gene ID
55757
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
HUGT2, UGCGL2, UGT2
Chromosome Location
Chr. 13: 95801580 - 96053482 on Build GRCh38
UniGene ID
Hs.193226
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020121.3 NP_064506.3 15 1786
XM_011521094.1 XP_011519396.1 15 1741
XM_011521095.1 XP_011519397.1 15 1741
XM_011521096.1 XP_011519398.1 15 1741
XM_011521097.1 XP_011519399.1 15 1741
XM_011521098.1 XP_011519400.1 8 758
XM_011521099.2 XP_011519401.1 7 675
XM_011521100.2 XP_011519402.1 15 1741
XM_011521102.2 XP_011519404.1 4 392
XM_011521103.2 XP_011519405.1 5 503
XM_017020662.1 XP_016876151.1 15 1741
XM_017020663.1 XP_016876152.1 16 1720
GenBank mRNA AF227906.2 - 15 1687
AK001735.1 - 15 1711
AL133051.1 - 12 1166
BC125233.1 - 15 1642 40124676

View Full Product Details