Gene Transcripts

Gene Symbol
PYROXD2
Entrez Gene ID
84795
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Gene Aliases
C10orf33, FP3420
Chromosome Location
Chr. 10: 98383565 - 98415221 on Build GRCh38
UniGene ID
Hs.238303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032709.2 NP_116098.2 5, 6 558
XM_011540293.2 XP_011538595.1 6, 7 1980
XM_017016835.1 XP_016872324.1 5, 6 519
XM_017016836.1 XP_016872325.1 5, 6 519
XM_017016837.1 XP_016872326.1 5, 6 519
XM_017016838.1 XP_016872327.1 5, 6 2754
XM_017016839.1 XP_016872328.1 3, 4 611
XM_017016840.1 XP_016872329.1 3, 4 465
XM_017016841.1 XP_016872330.1 2, 3 772
GenBank mRNA AK074429.1 - 5, 6 519
AK075265.1 - 5, 6 496
AK225637.1 - 5, 6 512
AK295699.1 - 5, 6 531
BC006131.1 - 5, 6 521 3626506

View Full Product Details