Gene Transcripts

Gene Symbol
ARHGAP19
Entrez Gene ID
84986
Gene Name
Rho GTPase activating protein 19
Gene Aliases
Chromosome Location
Chr. 10: 97222173 - 97292673 on Build GRCh38
UniGene ID
Hs.80305
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204300.1 NP_001191229.1 6 913
NM_001256423.1 NP_001243352.1 7 983
NM_032900.5 NP_116289.4 7 1000
GenBank mRNA AK074122.1 - 2 80
AK090447.1 - 8 1045
AK093316.1 - 5 631
AK093441.1 - 7 958
AK303055.1 - 7 961
BC113888.1 - 6 889 40082328
BC114490.1 - 7 976 40082327
BX648189.1 - 7 975
BX648194.1 - 4 443
CR749412.1 - 7 976
DQ338460.1 - 7 965

Gene Symbol
ARHGAP19-SLIT1
Entrez Gene ID
100533184
Gene Name
ARHGAP19-SLIT1 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 10: 97153042 - 97292673 on Build GRCh38
UniGene ID
Hs.80305
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037909.1 7 1000
GenBank mRNA AY336750.1 - 7 965

View Full Product Details