Gene Transcripts

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
Chromosome Location
Chr. 15: 82988297 - 83012298 on Build GRCh38
UniGene ID
Hs.352541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144597.2 NP_653198.2 3, 4 405
GenBank mRNA AK125586.1 - 2, 3 1491
AK312160.1 - 3, 4 379
BC019820.1 - 3, 4 370 4449390
KU178855.1 - 3, 4 290

View Full Product Details