Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 155018513 - 155033781 on Build GRCh38
UniGene ID
Hs.591491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144622.2 NP_653223.2 1 265
GenBank mRNA AK057496.1 - 1 265
BC111794.1 - 1 244
DB025590.1 - 1 265

View Full Product Details