Gene Transcripts

Gene Symbol
TEPSIN
Entrez Gene ID
146705
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Gene Aliases
C17orf56, ENTHD2
Chromosome Location
Chr. 17: 81228277 - 81239092 on Build GRCh38
UniGene ID
Hs.631761
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144679.2 NP_653280.1 4 325
XM_005257066.2 XP_005257123.1 5 919
XM_005257067.2 XP_005257124.1 4 325
XM_006721709.3 XP_006721772.1 4 332
XM_006721712.2 XP_006721775.1 4 763
XM_011524355.1 XP_011522657.1 5 925
XM_011524356.1 XP_011522658.1 5 925
XM_011524357.1 XP_011522659.1 4 953
XM_011524358.2 XP_011522660.1 5 854
XM_017024202.1 XP_016879691.1 5 1166
XM_017024203.1 XP_016879692.1 6 813
XM_017024204.1 XP_016879693.1 5 1366
GenBank mRNA AK056090.1 - 4 289
AK127221.1 - 2 1478
AK128728.1 - 5 1638
BC064483.1 - 4 286 5430837
DB122720.1 - 4 325

View Full Product Details