Gene Transcripts

Gene Symbol
LINC00479
Entrez Gene ID
150135
Gene Name
long intergenic non-protein coding RNA 479
Gene Aliases
C21orf129, PRED76
Chromosome Location
Chr. 21: 41711520 - 41715775 on Build GRCh38
UniGene ID
Hs.350679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027272.1 6 632
GenBank mRNA AK057397.1 - 6 632
BC128197.1 - 2 227
BC128198.1 - 6 653

View Full Product Details