Gene Transcripts

Gene Symbol
TDRP
Entrez Gene ID
157695
Gene Name
testis development related protein
Gene Aliases
C8orf42, Inm01, TDRP1, TDRP2
Chromosome Location
Chr. 8: 489790 - 545781 on Build GRCh38
UniGene ID
Hs.289293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256113.1 NP_001243042.1 3 437
NM_175075.4 NP_778250.2 4 605
XM_006716232.3 XP_006716295.1 3 585
XM_017013113.1 XP_016868602.1 3 919
GenBank mRNA AK126209.1 - 1 70
AY194292.2 - 3 423
BC136248.1 - 3 584 9020235
BC136249.1 - 3 584 9020236
BX648606.1 - 2 208
CN430635.1 - 3 416
FJ381685.1 - 3 429

View Full Product Details