Gene Transcripts

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
Chromosome Location
Chr. X: 80669488 - 80809736 on Build GRCh38
UniGene ID
Hs.147027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153252.4 NP_694984.4 35 4145
XM_005262113.3 XP_005262170.1 34 3997
XM_017029384.1 XP_016884873.1 31 3675
GenBank mRNA AK095887.1 - 18 2018
AY497046.1 - 35 4145
AY497047.1 - 32 4077
AY497048.1 - 35 4168
AY497049.1 - 34 4138
AY497050.1 - 35 4118
AY497051.1 - 32 3705
AY497052.1 - 37 4304
AY497053.1 - 33 3893
AY497054.1 - 32 3794
AY497055.1 - 35 4079
AY497056.1 - 32 4050
AY497057.1 - 31 3949
AY497058.1 - 31 3788
AY497059.1 - 33 4159
AY497060.1 - 29 3726
BC039508.2 - 7 618

View Full Product Details