Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000101.3 | NP_000092.2 | 4 | 341 | |
XM_011522905.2 | XP_011521207.1 | 4 | 336 | ||
GenBank mRNA | AK097127.1 | - | 4 | 244 | |
AK300049.1 | - | 4 | 245 | ||
AK309030.1 | - | 4 | 245 | ||
BC006465.1 | - | 4 | 297 | 3502788 | |
BT006861.1 | - | 4 | 270 | ||
BX109994.1 | - | 4 | 306 | ||
M21186.1 | - | 4 | 298 |