Gene Transcripts

Gene Symbol
GYPB
Entrez Gene ID
2994
Gene Name
glycophorin B (MNS blood group)
Gene Aliases
CD235b, GPB, GYP, MNS, PAS-3, SS
Chromosome Location
Chr. 4: 143995172 - 144021860 on Build GRCh38
UniGene ID
Hs.654368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304382.1 NP_001291311.1 4, 5 323
NM_002100.5 NP_002091.3 3, 4 243
XM_011531903.2 XP_011530205.1 3, 4 378
XM_011531904.2 XP_011530206.1 4, 5 2882
XM_011531905.2 XP_011530207.1 4, 5 530
XM_017008137.1 XP_016863626.1 3, 4 2577
GenBank mRNA AY950609.1 - 3, 4 185
AY950610.1 - 3, 4 185
AY950611.1 - 3, 4 185
AY950612.1 - 4, 5 278
BC069310.1 - 3, 4 235 7262104
BC121077.1 - 3, 4 219 40119713
BC121078.1 - 3, 4 219 40119716
EU338223.1 - 2, 3 123
EU338224.1 - 4, 5 346
EU338225.1 - 4, 5 315
EU338226.1 - 4, 5 280
EU338228.1 - 2, 3 123
EU338229.1 - 4, 5 346
EU338235.1 - 2, 3 123
EU338236.1 - 4, 5 346
EU338237.1 - 5, 6 439
EU338238.1 - 1, 2 84
J02982.1 - 3, 4 195
X08055.1 - 5, 6 321

View Full Product Details