Gene Transcripts

Gene Symbol
PMCHL1
Entrez Gene ID
5369
Gene Name
pro-melanin concentrating hormone like 1 (pseudogene)
Gene Aliases
Chromosome Location
Chr. 5: 22142352 - 22152272 on Build GRCh38
UniGene ID
Hs.247975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003921.1 4 525
GenBank mRNA AY008407.1 - 5 603
AY008408.1 - 5 607
AY008409.1 - 4 525
AY008410.1 - 4 563
AY008411.1 - 4 1784

Gene Symbol
PMCHL2
Entrez Gene ID
5370
Gene Name
pro-melanin concentrating hormone like 2 (pseudogene)
Gene Aliases
Chromosome Location
Chr. 5: 71375785 - 71385993 on Build GRCh38
UniGene ID
Hs.742169
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY008412.1 - 4 1783

View Full Product Details