Gene Transcripts

Gene Symbol
SLC39A4
Entrez Gene ID
55630
Gene Name
solute carrier family 39 member 4
Gene Aliases
AEZ, AWMS2, ZIP4
Chromosome Location
Chr. 8: 144412414 - 144416895 on Build GRCh38
UniGene ID
Hs.521934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017767.2 NP_060237.2 6 1485
NM_130849.3 NP_570901.2 7 1341
XM_006716599.1 XP_006716662.1 7 1320
GenBank mRNA AK000334.1 - 6 1465
AK000695.1 - 6 1400
AK025537.1 - 7 1335
AK290563.1 - 7 1289
BC062625.1 - 7 1321 5203858
KU178707.1 - 7 1235
KU178708.1 - 6 953

View Full Product Details