Gene Transcripts
- Gene Symbol
- SLC39A4
- Entrez Gene ID
- 55630
- Gene Name
- solute carrier family 39 member 4
- Gene Aliases
- AEZ, AWMS2, ZIP4
- Chromosome Location
- Chr. 8: 144412414 - 144416895 on Build GRCh38
- UniGene ID
- Hs.521934
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001280557.1 |
NP_001267486.1
|
1, 2 |
84 |
|
NM_017767.2 |
NP_060237.2
|
8, 9 |
1729 |
|
NM_130849.3 |
NP_570901.2
|
9, 10 |
1585 |
|
GenBank mRNA |
AK000334.1 |
- |
8, 9 |
1709 |
|
AK000489.1 |
- |
1, 2 |
79 |
|
AK000695.1 |
- |
8, 9 |
1644 |
|
AK025537.1 |
- |
9, 10 |
1579 |
|
AK056900.1 |
- |
1, 2 |
78 |
|
AK290563.1 |
- |
9, 10 |
1533 |
|
BC001688.2 |
- |
1, 2 |
56 |
|
BC062625.1 |
- |
9, 10 |
1565 |
5203858 |
KU178707.1 |
- |
9, 10 |
1479 |
|
KU178708.1 |
- |
8, 9 |
1197 |
|
View Full Product Details