Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
84266
Gene Name
alkB homolog 7
Gene Aliases
ABH7, SPATA11, UNQ6002
Chromosome Location
Chr. 19: 6372433 - 6375250 on Build GRCh38
UniGene ID
Hs.111099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032306.3 NP_115682.1 4 935
XM_005259658.4 XP_005259715.1 4 511
XM_017027355.1 XP_016882844.1 3 792
GenBank mRNA AK223336.1 - 4 558
AK311956.1 - 4 574
AY358858.1 - 4 935
AY427650.2 - 4 935
BC004393.1 - 4 590 3635540
BM271861.1 - 1 428

View Full Product Details