Gene Transcripts

Gene Symbol
C2orf76
Entrez Gene ID
130355
Gene Name
chromosome 2 open reading frame 76
Gene Aliases
AIM29
Chromosome Location
Chr. 2: 119281472 - 119366879 on Build GRCh38
UniGene ID
Hs.99488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017927.3 NP_001017927.2 6 878
NM_001322329.1 NP_001309258.1 6 799
NM_001322330.1 NP_001309259.1 5 675
NM_001322331.1 NP_001309260.1 5 328
NM_001322332.1 NP_001309261.1 4 277
XM_017003354.1 XP_016858843.1 5 377
GenBank mRNA AW015242.1 - 2 375
BC047365.1 - 6 799 5189916
BC062434.1 - 4 310
BC105805.1 - 5 332 6496685
BC126395.1 - 5 615 8992111
BC126397.1 - 6 818 8992113
BC144012.1 - 6 818 9052531
BC171737.1 - 5 615 9052532

View Full Product Details