Gene Transcripts

Gene Symbol
SAMD10
Entrez Gene ID
140700
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
C20orf136, dJ591C20, dJ591C20.7
Chromosome Location
Chr. 20: 63974113 - 63980788 on Build GRCh38
UniGene ID
Hs.27189
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080621.4 NP_542188.1 3 550
XM_005260199.4 XP_005260256.1 5 1687
XM_006723705.3 XP_006723768.1 4 449
XM_011528565.2 XP_011526867.1 4 1319
XM_017027671.1 XP_016883160.1 4 1567
GenBank mRNA BC067362.1 - 3 547

View Full Product Details