Gene Transcripts

Gene Symbol
ADAM5
Entrez Gene ID
255926
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases
ADAM5P, TMDCII
Chromosome Location
Chr. 8: 39314663 - 39417226 on Build GRCh38
UniGene ID
Hs.369765
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001448.1 11, 12 1288
GenBank mRNA AJ132821.1 - 10, 11 1222
AJ132822.1 - 10, 11 1104
AJ132823.1 - 9, 10 1038
AJ132827.1 - 1, 2 218
BC047448.1 - 11, 12 1422
BC067864.1 - 9, 10 1081

View Full Product Details