Gene Transcripts

Gene Symbol
FAM89A
Entrez Gene ID
375061
Gene Name
family with sequence similarity 89 member A
Gene Aliases
C1orf153
Chromosome Location
Chr. 1: 231018958 - 231040249 on Build GRCh38
UniGene ID
Hs.38516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198552.2 NP_940954.1 2 434
GenBank mRNA BC009447.1 - 2 424

View Full Product Details