Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025197.1 | NP_001020368.1 | 3 | 354 | |
NM_001025199.1 | NP_001020370.1 | 3 | 414 | ||
NM_004000.2 | NP_003991.2 | 4 | 384 | ||
GenBank mRNA | AK223316.1 | - | 3 | 294 | |
AK298466.1 | - | 3 | 305 | ||
BC011460.1 | - | 4 | 322 | 4214121 | |
BG698401.1 | - | 4 | 384 | ||
BM787868.1 | - | 3 | 404 | ||
BT006767.1 | - | 4 | 313 | ||
U49835.1 | - | 4 | 333 | ||
U58514.1 | - | 4 | 349 | ||
U58515.1 | - | 3 | 415 |