Gene Transcripts

Gene Symbol
SLC9B1
Entrez Gene ID
150159
Gene Name
solute carrier family 9 member B1
Gene Aliases
NHA1, NHEDC1
Chromosome Location
Chr. 4: 102885048 - 103019739 on Build GRCh38
UniGene ID
Hs.666728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100874.2 NP_001094344.1 3, 4 364
NM_139173.3 NP_631912.2 3, 4 364
NR_047513.1 3, 4 364
XM_006714093.3 XP_006714156.1 4, 5 383
XM_011531622.1 XP_011529924.1 4, 5 347
XM_011531623.1 XP_011529925.1 3, 4 351
GenBank mRNA AF447585.1 - 3, 4 344
AK313592.1 - 3, 4 291
AY461581.1 - 3, 4 312
BC046636.1 - 3, 4 288
BC110794.1 - 3, 4 248 5745227
BC132712.1 - 3, 4 291 40146734
BC136966.1 - 3, 4 291 9020958
DQ003059.1 - 3, 4 344

View Full Product Details