Gene Transcripts

Gene Symbol
FAM162B
Entrez Gene ID
221303
Gene Name
family with sequence similarity 162 member B
Gene Aliases
C6orf189, bA86F4.2
Chromosome Location
Chr. 6: 116752197 - 116765723 on Build GRCh38
UniGene ID
Hs.126712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085480.2 NP_001078949.1 3 500
GenBank mRNA BC038997.1 - 3 499

View Full Product Details