Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139021.2 | NP_620590.2 | 11 | 1170 | |
XM_011516925.2 | XP_011515227.1 | 11 | 2008 | ||
XM_011516926.2 | XP_011515228.1 | 11 | 1259 | ||
XM_011516927.2 | XP_011515229.1 | 11 | 1951 | ||
XM_017013218.1 | XP_016868707.1 | 10 | 1915 | ||
GenBank mRNA | AY065978.1 | - | 11 | 1162 | |
BC028034.1 | - | 9 | 1527 | 5242481 |