Gene Transcripts

Gene Symbol
SOWAHB
Entrez Gene ID
345079
Gene Name
sosondowah ankyrin repeat domain family member B
Gene Aliases
ANKRD56
Chromosome Location
Chr. 4: 76894929 - 76897849 on Build GRCh38
UniGene ID
Hs.257292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029870.1 NP_001025041.1 1 2128
GenBank mRNA BC137241.1 - 1 2205
BC137255.1 - 1 2205 9021252

View Full Product Details