Gene Transcripts

Gene Symbol
NDUFV2
Entrez Gene ID
4729
Gene Name
NADH:ubiquinone oxidoreductase core subunit V2
Gene Aliases
CI-24k
Chromosome Location
Chr. 18: 9102623 - 9134345 on Build GRCh38
UniGene ID
Hs.464572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021074.4 NP_066552.2 5 544
XM_017025782.1 XP_016881271.1 5 734
GenBank mRNA AF147367.1 - 1 19
AK291865.1 - 5 440
AK312129.1 - 5 462
AW051350.1 - 4 390
BC001632.1 - 5 454 3536345
BC017487.1 - 5 452 3537815
BC071689.1 - 5 434 4372795
BG742948.1 - 5 544
CR456928.1 - 5 430
M22538.1 - 5 448

View Full Product Details