Gene Transcripts
- Gene Symbol
- CNTLN
- Entrez Gene ID
- 54875
- Gene Name
- centlein
- Gene Aliases
- C9orf101, C9orf39, bA340N12.1
- Chromosome Location
- Chr. 9: 17134991 - 17510019 on Build GRCh38
- UniGene ID
- Hs.435381
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114395.2 |
NP_001107867.1
|
2, 3 |
514 |
|
NM_017738.3 |
NP_060208.2
|
2, 3 |
514 |
|
XM_005251492.1 |
XP_005251549.1
|
2, 3 |
514 |
|
XM_006716793.3 |
XP_006716856.1
|
2, 3 |
312 |
|
XM_011517941.2 |
XP_011516243.1
|
2, 3 |
514 |
|
XM_017014839.1 |
XP_016870328.1
|
2, 3 |
514 |
|
XM_017014840.1 |
XP_016870329.1
|
2, 3 |
514 |
|
XM_017014841.1 |
XP_016870330.1
|
2, 3 |
514 |
|
XM_017014842.1 |
XP_016870331.1
|
2, 3 |
514 |
|
XM_017014843.1 |
XP_016870332.1
|
2, 3 |
514 |
|
GenBank mRNA |
AK098502.1 |
- |
2, 3 |
465 |
|
AK308837.1 |
- |
2, 3 |
469 |
|
BX647069.1 |
- |
2, 3 |
472 |
|
DB079230.1 |
- |
2, 3 |
515 |
|
View Full Product Details