Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145725.1 | NP_001139197.1 | 5, 6 | 496 | |
NM_017816.2 | NP_060286.1 | 5, 6 | 594 | ||
XM_011513505.1 | XP_011511807.1 | 5, 6 | 896 | ||
XM_011513506.2 | XP_011511808.1 | 4, 5 | 527 | ||
GenBank mRNA | AK000432.1 | - | 5, 6 | 541 | |
AK290750.1 | - | 5, 6 | 571 | ||
AL136750.1 | - | 5, 6 | 496 | ||
BC015796.2 | - | 5, 6 | 556 | 3911275 | |
CR533548.1 | - | 3, 4 | 353 |