Gene Transcripts

Gene Symbol
FSCN1
Entrez Gene ID
6624
Gene Name
fascin actin-bundling protein 1
Gene Aliases
FAN1, HSN, SNL, p55
Chromosome Location
Chr. 7: 5592805 - 5606656 on Build GRCh38
UniGene ID
Hs.118400
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003088.3 NP_003079.1 1 219
GenBank mRNA AK095254.1 - 1 216
AK316607.1 - 1 214
BC000521.2 - 1 110 2961047
BC006304.2 - 1 201 4100996
BC007539.1 - 1 110
BC007948.2 - 1 198 4299524
BC007988.2 - 1 110 2961047
BI546742.1 - 1 218
BT006636.1 - 1 87
U03057.1 - 1 198
U09873.1 - 1 132

View Full Product Details