Gene Transcripts

Gene Symbol
CALML3
Entrez Gene ID
810
Gene Name
calmodulin like 3
Gene Aliases
CLP
Chromosome Location
Chr. 10: 5524889 - 5526268 on Build GRCh38
UniGene ID
Hs.239600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005185.3 NP_005176.1 1 365
GenBank mRNA AK313934.1 - 1 293
BC031889.1 - 1 273 4777524
M36707.1 - 1 1233
M58026.1 - 1 293

View Full Product Details