Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85659697-85660198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBZF
Entrez Gene ID
58487
Gene Name
CREB/ATF bZIP transcription factor
Gene Aliases
SMILE, ZF
UniGene
Hs.535319
Gene Chromosome Location
Chr.11: 85657564-85665148 on Build GRCh38
Transcripts
NM_001039618.2 NR_028024.1 NR_028025.1 NR_028026.1 NR_028027.1 XM_006718642.3 XM_011545195.2 XM_017018086.1 XM_017018087.1 XM_017018088.1 XM_017018089.1 XM_017018090.1 XM_017018091.1 XM_017018092.1

SNP Detail

SNP ID Chromosome Location
rs1041035672 Chr.11: 85659804-85659804 on Build GRCh38
rs79279804 Chr.11: 85660085-85660085 on Build GRCh38
rs183448595 Chr.11: 85659786-85659786 on Build GRCh38
rs1006079229 Chr.11: 85659906-85659906 on Build GRCh38
rs144522088 Chr.11: 85659762-85659764 on Build GRCh38
rs756319224 Chr.11: 85660120-85660120 on Build GRCh38
rs1024758138 Chr.11: 85659764-85659764 on Build GRCh38
rs905625931 Chr.11: 85660014-85660014 on Build GRCh38
rs777157310 Chr.11: 85659927-85659928 on Build GRCh38
rs148456519 Chr.11: 85660105-85660105 on Build GRCh38
rs947614604 Chr.11: 85659727-85659727 on Build GRCh38
rs887415298 Chr.11: 85659729-85659729 on Build GRCh38
rs1002507229 Chr.11: 85660049-85660049 on Build GRCh38
rs976861972 Chr.11: 85659999-85659999 on Build GRCh38
rs3168151 Chr.11: 85659899-85659899 on Build GRCh38
rs977962266 Chr.11: 85659812-85659812 on Build GRCh38
rs780270522 Chr.11: 85660153-85660153 on Build GRCh38
rs1023114418 Chr.11: 85659953-85659953 on Build GRCh38
rs1005897517 Chr.11: 85659747-85659747 on Build GRCh38
rs190873798 Chr.11: 85659749-85659749 on Build GRCh38
rs537045811 Chr.11: 85660003-85660003 on Build GRCh38
rs941650752 Chr.11: 85660109-85660109 on Build GRCh38
rs918740671 Chr.11: 85660011-85660012 on Build GRCh38
rs375730954 Chr.11: 85659748-85659748 on Build GRCh38
rs1054793577 Chr.11: 85660138-85660141 on Build GRCh38
rs796563728 Chr.11: 85659759-85659761 on Build GRCh38
rs867556822 Chr.11: 85659988-85659988 on Build GRCh38
rs146717028 Chr.11: 85659902-85659902 on Build GRCh38
rs888711183 Chr.11: 85660100-85660103 on Build GRCh38
rs944134016 Chr.11: 85659967-85659967 on Build GRCh38
rs1048700855 Chr.11: 85659723-85659724 on Build GRCh38
rs925223707 Chr.11: 85659828-85659828 on Build GRCh38
rs929226164 Chr.11: 85659834-85659834 on Build GRCh38
rs930145374 Chr.11: 85660049-85660049 on Build GRCh38
rs901732296 Chr.11: 85659815-85659815 on Build GRCh38
rs911333174 Chr.11: 85659762-85659762 on Build GRCh38
rs911257428 Chr.11: 85659947-85659947 on Build GRCh38

View Full Product Details