Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2449029-2449531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1C
Entrez Gene ID
775
Gene Name
calcium voltage-gated channel subunit alpha1 C
Gene Aliases
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
UniGene
Hs.118262
Gene Chromosome Location
Chr.12: 1969677-2697949 on Build GRCh38
Transcripts
NM_000719.6 NM_001129827.1 NM_001129829.1 NM_001129830.2 NM_001129831.1 NM_001129832.1 NM_001129833.1 NM_001129834.1 NM_001129835.1 NM_001129836.1 NM_001129837.1 NM_001129838.1 NM_001129839.1 NM_001129840.1 NM_001129841.1 NM_001129842.1 NM_001129843.1 NM_001129844.1 NM_001129846.1 NM_001167623.1 NM_001167624.2 NM_001167625.1 NM_199460.3 XM_006719017.2 XM_011521020.2 XM_011521023.2 XM_017019926.1 XM_017019927.1 XM_017019928.1 XM_017019929.1 XM_017019930.1 XM_017019931.1 XM_017019932.1 XM_017019933.1 XM_017019934.1 XM_017019935.1 XM_017019936.1 XM_017019937.1 XM_017019938.1 XM_017019939.1 XM_017019940.1 XM_017019941.1 XM_017019942.1 XM_017019943.1 XM_017019944.1 XM_017019945.1 XM_017019946.1 XM_017019947.1 XM_017019948.1 XM_017019949.1 XM_017019950.1 XM_017019951.1 XM_017019952.1 XM_017019953.1 XM_017019954.1 XM_017019955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2202594 c.608T>C p.V203A Chr.12: 2449106-2449106 on Build GRCh38
7202267 c.616G>A p.G206R Chr.12: 2449114-2449114 on Build GRCh38
2202588 c.537C>T p.I179I Chr.12: 2449035-2449035 on Build GRCh38
4780064 c.669C>T p.N223N Chr.12: 2449077-2449077 on Build GRCh38
3739577 c.617+8T>A p.? Chr.12: 2449123-2449123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969789637 Chr.12: 2449073-2449073 on Build GRCh38
rs886049202 Chr.12: 2449065-2449065 on Build GRCh38
rs111778211 Chr.12: 2449133-2449133 on Build GRCh38
rs538997856 Chr.12: 2449060-2449060 on Build GRCh38
rs989604172 Chr.12: 2449451-2449451 on Build GRCh38
rs372770722 Chr.12: 2449068-2449068 on Build GRCh38
rs1036463390 Chr.12: 2449354-2449354 on Build GRCh38
rs375602020 Chr.12: 2449445-2449445 on Build GRCh38
rs566888241 Chr.12: 2449319-2449319 on Build GRCh38
rs763452803 Chr.12: 2449196-2449196 on Build GRCh38
rs960804354 Chr.12: 2449328-2449328 on Build GRCh38
rs953638150 Chr.12: 2449456-2449456 on Build GRCh38
rs1031390595 Chr.12: 2449429-2449429 on Build GRCh38
rs979255353 Chr.12: 2449380-2449380 on Build GRCh38
rs971155109 Chr.12: 2449225-2449225 on Build GRCh38
rs148166373 Chr.12: 2449425-2449425 on Build GRCh38
rs913717979 Chr.12: 2449162-2449162 on Build GRCh38
rs749405526 Chr.12: 2449141-2449141 on Build GRCh38
rs566358985 Chr.12: 2449259-2449259 on Build GRCh38
rs386759537 Chr.12: 2449132-2449134 on Build GRCh38
rs550267364 Chr.12: 2449126-2449126 on Build GRCh38
rs897631336 Chr.12: 2449376-2449376 on Build GRCh38
rs991165917 Chr.12: 2449205-2449205 on Build GRCh38
rs995137039 Chr.12: 2449283-2449283 on Build GRCh38
rs4765682 Chr.12: 2449401-2449401 on Build GRCh38
rs928040016 Chr.12: 2449126-2449129 on Build GRCh38
rs548482522 Chr.12: 2449313-2449313 on Build GRCh38
rs766754214 Chr.12: 2449257-2449257 on Build GRCh38
rs879015400 Chr.12: 2449087-2449087 on Build GRCh38
rs530130732 Chr.12: 2449119-2449119 on Build GRCh38
rs995420910 Chr.12: 2449454-2449454 on Build GRCh38
rs4765933 Chr.12: 2449309-2449309 on Build GRCh38
rs767270982 Chr.12: 2449062-2449062 on Build GRCh38
rs994677032 Chr.12: 2449397-2449397 on Build GRCh38
rs557373646 Chr.12: 2449169-2449169 on Build GRCh38
rs913774837 Chr.12: 2449346-2449346 on Build GRCh38
rs75588355 Chr.12: 2449310-2449310 on Build GRCh38
rs1544515 Chr.12: 2449132-2449132 on Build GRCh38
rs890476765 Chr.12: 2449334-2449334 on Build GRCh38
rs986807009 Chr.12: 2449488-2449488 on Build GRCh38
rs1544516 Chr.12: 2449134-2449134 on Build GRCh38
rs958578529 Chr.12: 2449174-2449174 on Build GRCh38
rs977596664 Chr.12: 2449238-2449238 on Build GRCh38
rs1050690866 Chr.12: 2449213-2449213 on Build GRCh38
rs1002438431 Chr.12: 2449481-2449481 on Build GRCh38
rs912218878 Chr.12: 2449424-2449424 on Build GRCh38
rs774539191 Chr.12: 2449164-2449164 on Build GRCh38
rs1027583006 Chr.12: 2449168-2449168 on Build GRCh38
rs752657132 Chr.12: 2449452-2449452 on Build GRCh38
rs74823149 Chr.12: 2449312-2449312 on Build GRCh38
rs759633434 Chr.12: 2449335-2449335 on Build GRCh38
rs552535781 Chr.12: 2449256-2449256 on Build GRCh38
rs771078944 Chr.12: 2449161-2449161 on Build GRCh38
rs935515055 Chr.12: 2449273-2449273 on Build GRCh38
rs78824737 Chr.12: 2449311-2449311 on Build GRCh38
rs1024424233 Chr.12: 2449206-2449206 on Build GRCh38
rs747320984 Chr.12: 2449113-2449113 on Build GRCh38
rs572213192 Chr.12: 2449441-2449441 on Build GRCh38
rs971487387 Chr.12: 2449418-2449418 on Build GRCh38
rs905460214 Chr.12: 2449480-2449480 on Build GRCh38
rs962347616 Chr.12: 2449065-2449065 on Build GRCh38
rs1023952578 Chr.12: 2449417-2449417 on Build GRCh38
rs569448710 Chr.12: 2449407-2449407 on Build GRCh38
rs555783569 Chr.12: 2449402-2449402 on Build GRCh38
rs759335182 Chr.12: 2449057-2449057 on Build GRCh38
rs377571606 Chr.12: 2449086-2449086 on Build GRCh38
rs538819011 Chr.12: 2449408-2449408 on Build GRCh38
rs778711543 Chr.12: 2449092-2449092 on Build GRCh38
rs897983159 Chr.12: 2449266-2449266 on Build GRCh38
rs756929253 Chr.12: 2449084-2449084 on Build GRCh38
rs1027530636 Chr.12: 2449400-2449400 on Build GRCh38
rs558520264 Chr.12: 2449431-2449431 on Build GRCh38
rs774798005 Chr.12: 2449285-2449286 on Build GRCh38
rs921152071 Chr.12: 2449196-2449196 on Build GRCh38
rs779735913 Chr.12: 2449079-2449079 on Build GRCh38
rs753418562 Chr.12: 2449078-2449078 on Build GRCh38
rs748658616 Chr.12: 2449130-2449130 on Build GRCh38
rs192177703 Chr.12: 2449373-2449373 on Build GRCh38
rs561224137 Chr.12: 2449077-2449077 on Build GRCh38
rs756042537 Chr.12: 2449072-2449072 on Build GRCh38
rs946683884 Chr.12: 2449170-2449170 on Build GRCh38
rs1050437536 Chr.12: 2449330-2449330 on Build GRCh38
rs960688400 Chr.12: 2449495-2449495 on Build GRCh38

View Full Product Details