Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56237066-56237571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A5
Entrez Gene ID
283375
Gene Name
solute carrier family 39 member 5
Gene Aliases
LZT-Hs7, MYP24, ZIP5
UniGene
Hs.591018
Gene Chromosome Location
Chr.12: 56230036-56237846 on Build GRCh38
Transcripts
NM_001135195.1 NM_173596.2 XM_005268803.1 XM_011538198.1 XM_011538199.1 XM_011538200.1 XM_011538201.1 XM_017019185.1 XM_017019186.1 XM_017019187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7256559 c.1452C>G p.V484V Chr.12: 56237313-56237313 on Build GRCh38
4854299 c.1295T>C p.F432S Chr.12: 56237156-56237156 on Build GRCh38
3727587 c.1448G>A p.G483E Chr.12: 56237309-56237309 on Build GRCh38
1317984 c.1392G>T p.V464V Chr.12: 56237253-56237253 on Build GRCh38
5841531 c.1427G>A p.W476* Chr.12: 56237288-56237288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777732265 Chr.12: 56237369-56237369 on Build GRCh38
rs776558122 Chr.12: 56237217-56237217 on Build GRCh38
rs770937674 Chr.12: 56237309-56237309 on Build GRCh38
rs1032265879 Chr.12: 56237159-56237159 on Build GRCh38
rs778228302 Chr.12: 56237231-56237232 on Build GRCh38
rs773027157 Chr.12: 56237523-56237523 on Build GRCh38
rs531957758 Chr.12: 56237144-56237144 on Build GRCh38
rs781230054 Chr.12: 56237388-56237388 on Build GRCh38
rs753590060 Chr.12: 56237129-56237129 on Build GRCh38
rs779311053 Chr.12: 56237345-56237345 on Build GRCh38
rs531965517 Chr.12: 56237495-56237495 on Build GRCh38
rs199593141 Chr.12: 56237149-56237149 on Build GRCh38
rs564511364 Chr.12: 56237491-56237491 on Build GRCh38
rs998398686 Chr.12: 56237487-56237488 on Build GRCh38
rs1006809050 Chr.12: 56237524-56237524 on Build GRCh38
rs768764585 Chr.12: 56237111-56237111 on Build GRCh38
rs201955617 Chr.12: 56237549-56237549 on Build GRCh38
rs984565714 Chr.12: 56237291-56237291 on Build GRCh38
rs377171641 Chr.12: 56237356-56237356 on Build GRCh38
rs372580129 Chr.12: 56237286-56237286 on Build GRCh38
rs373073769 Chr.12: 56237211-56237211 on Build GRCh38
rs768850888 Chr.12: 56237191-56237191 on Build GRCh38
rs369774679 Chr.12: 56237249-56237249 on Build GRCh38
rs746981581 Chr.12: 56237303-56237303 on Build GRCh38
rs780454338 Chr.12: 56237250-56237250 on Build GRCh38
rs146756761 Chr.12: 56237181-56237181 on Build GRCh38
rs560052706 Chr.12: 56237269-56237269 on Build GRCh38
rs929019455 Chr.12: 56237289-56237289 on Build GRCh38
rs769753149 Chr.12: 56237539-56237539 on Build GRCh38
rs891850439 Chr.12: 56237179-56237179 on Build GRCh38
rs773633117 Chr.12: 56237151-56237152 on Build GRCh38
rs1054109565 Chr.12: 56237088-56237088 on Build GRCh38
rs907417543 Chr.12: 56237386-56237386 on Build GRCh38
rs766805438 Chr.12: 56237337-56237337 on Build GRCh38
rs752898287 Chr.12: 56237347-56237347 on Build GRCh38
rs76216511 Chr.12: 56237201-56237201 on Build GRCh38
rs762636627 Chr.12: 56237328-56237328 on Build GRCh38
rs368515048 Chr.12: 56237105-56237105 on Build GRCh38
rs763578338 Chr.12: 56237330-56237330 on Build GRCh38
rs145213363 Chr.12: 56237195-56237195 on Build GRCh38
rs578087997 Chr.12: 56237343-56237343 on Build GRCh38
rs554289471 Chr.12: 56237403-56237403 on Build GRCh38
rs374778265 Chr.12: 56237114-56237114 on Build GRCh38
rs775351499 Chr.12: 56237540-56237540 on Build GRCh38
rs766148953 Chr.12: 56237122-56237122 on Build GRCh38
rs758095176 Chr.12: 56237322-56237323 on Build GRCh38
rs771025597 Chr.12: 56237387-56237387 on Build GRCh38
rs113388846 Chr.12: 56237429-56237429 on Build GRCh38
rs763639581 Chr.12: 56237235-56237235 on Build GRCh38
rs754143279 Chr.12: 56237251-56237251 on Build GRCh38
rs772075445 Chr.12: 56237198-56237198 on Build GRCh38
rs369482324 Chr.12: 56237340-56237340 on Build GRCh38
rs761183100 Chr.12: 56237335-56237335 on Build GRCh38
rs777873914 Chr.12: 56237299-56237299 on Build GRCh38
rs762152137 Chr.12: 56237339-56237339 on Build GRCh38
rs143299854 Chr.12: 56237218-56237218 on Build GRCh38
rs772091180 Chr.12: 56237288-56237288 on Build GRCh38
rs572486882 Chr.12: 56237415-56237415 on Build GRCh38
rs188130856 Chr.12: 56237327-56237327 on Build GRCh38
rs549345170 Chr.12: 56237089-56237089 on Build GRCh38
rs770088607 Chr.12: 56237102-56237102 on Build GRCh38
rs140453919 Chr.12: 56237188-56237188 on Build GRCh38
rs150400071 Chr.12: 56237189-56237189 on Build GRCh38
rs201599048 Chr.12: 56237151-56237151 on Build GRCh38
rs754643129 Chr.12: 56237140-56237140 on Build GRCh38
rs370427401 Chr.12: 56237106-56237106 on Build GRCh38
rs745473878 Chr.12: 56237232-56237232 on Build GRCh38
rs372737605 Chr.12: 56237349-56237349 on Build GRCh38
rs938946594 Chr.12: 56237418-56237418 on Build GRCh38
rs888234271 Chr.12: 56237434-56237434 on Build GRCh38
rs547030232 Chr.12: 56237544-56237544 on Build GRCh38
rs776576355 Chr.12: 56237310-56237310 on Build GRCh38
rs1005254235 Chr.12: 56237134-56237134 on Build GRCh38
rs750934470 Chr.12: 56237239-56237239 on Build GRCh38
rs766313655 Chr.12: 56237212-56237212 on Build GRCh38
rs374099812 Chr.12: 56237436-56237436 on Build GRCh38
rs779065616 Chr.12: 56237255-56237255 on Build GRCh38
rs769594905 Chr.12: 56237325-56237325 on Build GRCh38
rs764519283 Chr.12: 56237341-56237344 on Build GRCh38
rs143219697 Chr.12: 56237230-56237230 on Build GRCh38
rs200932335 Chr.12: 56237145-56237145 on Build GRCh38
rs528934105 Chr.12: 56237166-56237166 on Build GRCh38
rs937867192 Chr.12: 56237407-56237407 on Build GRCh38
rs889783987 Chr.12: 56237501-56237501 on Build GRCh38
rs755195261 Chr.12: 56237253-56237253 on Build GRCh38
rs759368291 Chr.12: 56237316-56237316 on Build GRCh38
rs745779162 Chr.12: 56237392-56237392 on Build GRCh38
rs919101222 Chr.12: 56237237-56237237 on Build GRCh38
rs891983614 Chr.12: 56237444-56237444 on Build GRCh38
rs117751186 Chr.12: 56237445-56237445 on Build GRCh38
rs139208152 Chr.12: 56237228-56237228 on Build GRCh38
rs757799654 Chr.12: 56237232-56237232 on Build GRCh38
rs141805625 Chr.12: 56237461-56237461 on Build GRCh38
rs892416039 Chr.12: 56237095-56237095 on Build GRCh38
rs747048259 Chr.12: 56237370-56237370 on Build GRCh38
rs199870707 Chr.12: 56237192-56237192 on Build GRCh38
rs368085387 Chr.12: 56237118-56237118 on Build GRCh38
rs774410618 Chr.12: 56237112-56237112 on Build GRCh38
rs749351237 Chr.12: 56237120-56237120 on Build GRCh38
rs200013030 Chr.12: 56237331-56237331 on Build GRCh38
rs927785743 Chr.12: 56237358-56237358 on Build GRCh38

View Full Product Details