Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56642595-56643107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5B
Entrez Gene ID
506
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
ATPMB, ATPSB, HEL-S-271
UniGene
Hs.406510
Gene Chromosome Location
Chr.12: 56638175-56646068 on Build GRCh38
Transcripts
NM_001686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1512589 c.885C>T p.D295D Chr.12: 56642739-56642739 on Build GRCh38
3376318 c.898G>C p.D300H Chr.12: 56642726-56642726 on Build GRCh38
4633702 c.854C>A p.T285N Chr.12: 56642770-56642770 on Build GRCh38
3968337 c.936C>G p.T312T Chr.12: 56642688-56642688 on Build GRCh38
4513387 c.936C>T p.T312T Chr.12: 56642688-56642688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044365429 Chr.12: 56643001-56643001 on Build GRCh38
rs749480463 Chr.12: 56642627-56642627 on Build GRCh38
rs796429610 Chr.12: 56642985-56642986 on Build GRCh38
rs1044985091 Chr.12: 56642793-56642793 on Build GRCh38
rs750895114 Chr.12: 56642727-56642727 on Build GRCh38
rs780433436 Chr.12: 56642770-56642770 on Build GRCh38
rs369008512 Chr.12: 56642662-56642662 on Build GRCh38
rs749568756 Chr.12: 56642784-56642784 on Build GRCh38
rs200074324 Chr.12: 56642935-56642935 on Build GRCh38
rs768525884 Chr.12: 56643018-56643018 on Build GRCh38
rs201302572 Chr.12: 56642625-56642625 on Build GRCh38
rs745687845 Chr.12: 56642968-56642968 on Build GRCh38
rs1019755484 Chr.12: 56642893-56642893 on Build GRCh38
rs747026741 Chr.12: 56642838-56642838 on Build GRCh38
rs752204083 Chr.12: 56642716-56642716 on Build GRCh38
rs552591623 Chr.12: 56642904-56642904 on Build GRCh38
rs961266855 Chr.12: 56642997-56642997 on Build GRCh38
rs760397757 Chr.12: 56642652-56642652 on Build GRCh38
rs189451411 Chr.12: 56642630-56642630 on Build GRCh38
rs895828024 Chr.12: 56642676-56642676 on Build GRCh38
rs748238894 Chr.12: 56642846-56642846 on Build GRCh38
rs752290085 Chr.12: 56642872-56642872 on Build GRCh38
rs1028066248 Chr.12: 56642922-56642922 on Build GRCh38
rs542763979 Chr.12: 56642699-56642699 on Build GRCh38
rs756694263 Chr.12: 56642840-56642840 on Build GRCh38
rs11542644 Chr.12: 56642680-56642680 on Build GRCh38
rs762473573 Chr.12: 56642876-56642876 on Build GRCh38
rs140513120 Chr.12: 56642718-56642718 on Build GRCh38
rs200319393 Chr.12: 56642744-56642744 on Build GRCh38
rs536472466 Chr.12: 56643005-56643005 on Build GRCh38
rs377659522 Chr.12: 56642854-56642854 on Build GRCh38
rs775213376 Chr.12: 56642983-56642983 on Build GRCh38
rs916031834 Chr.12: 56642756-56642756 on Build GRCh38
rs768985218 Chr.12: 56642826-56642826 on Build GRCh38
rs778003361 Chr.12: 56642723-56642723 on Build GRCh38
rs1048274030 Chr.12: 56643029-56643029 on Build GRCh38
rs764295123 Chr.12: 56642757-56642758 on Build GRCh38
rs201322205 Chr.12: 56642742-56642742 on Build GRCh38
rs373472201 Chr.12: 56642634-56642634 on Build GRCh38
rs759255437 Chr.12: 56642861-56642861 on Build GRCh38
rs886961459 Chr.12: 56643049-56643049 on Build GRCh38
rs760801755 Chr.12: 56642663-56642664 on Build GRCh38
rs762449948 Chr.12: 56642867-56642869 on Build GRCh38
rs779338647 Chr.12: 56642631-56642631 on Build GRCh38
rs565725846 Chr.12: 56642964-56642964 on Build GRCh38
rs2272314 Chr.12: 56642916-56642916 on Build GRCh38
rs78744402 Chr.12: 56642986-56642986 on Build GRCh38
rs948788024 Chr.12: 56642995-56642995 on Build GRCh38
rs535120539 Chr.12: 56643013-56643013 on Build GRCh38
rs532126338 Chr.12: 56642911-56642911 on Build GRCh38
rs372963745 Chr.12: 56642844-56642844 on Build GRCh38
rs764789281 Chr.12: 56642700-56642700 on Build GRCh38
rs180693950 Chr.12: 56642912-56642912 on Build GRCh38
rs371036989 Chr.12: 56642858-56642858 on Build GRCh38
rs375233620 Chr.12: 56642626-56642626 on Build GRCh38
rs769605471 Chr.12: 56642973-56642973 on Build GRCh38
rs139524118 Chr.12: 56642903-56642903 on Build GRCh38
rs773130792 Chr.12: 56642635-56642635 on Build GRCh38
rs886901357 Chr.12: 56642866-56642866 on Build GRCh38
rs1042001 Chr.12: 56642804-56642804 on Build GRCh38
rs758934230 Chr.12: 56642717-56642717 on Build GRCh38
rs753476589 Chr.12: 56642665-56642665 on Build GRCh38
rs867442552 Chr.12: 56642753-56642753 on Build GRCh38
rs377383488 Chr.12: 56642852-56642852 on Build GRCh38
rs780807358 Chr.12: 56642954-56642954 on Build GRCh38
rs764878934 Chr.12: 56642868-56642868 on Build GRCh38
rs568255847 Chr.12: 56642986-56642988 on Build GRCh38
rs754375538 Chr.12: 56642865-56642865 on Build GRCh38
rs776399822 Chr.12: 56642860-56642860 on Build GRCh38
rs1800641 Chr.12: 56643070-56643070 on Build GRCh38
rs375196006 Chr.12: 56642853-56642853 on Build GRCh38

View Full Product Details