Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67649691-67650191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
UniGene
Hs.173135
Gene Chromosome Location
Chr.12: 67648732-67665405 on Build GRCh38
Transcripts
NM_003583.3 NM_006482.2 XM_017020032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4147413 c.148A>C p.I50L Chr.12: 67649895-67649895 on Build GRCh38
4147412 c.142T>C p.S48P Chr.12: 67649889-67649889 on Build GRCh38
3464446 c.57T>A p.G19G Chr.12: 67649804-67649804 on Build GRCh38
5549229 c.69C>G p.A23A Chr.12: 67649816-67649816 on Build GRCh38
6462851 c.51C>T p.G17G Chr.12: 67649798-67649798 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372712630 Chr.12: 67649758-67649758 on Build GRCh38
rs571967830 Chr.12: 67649856-67649856 on Build GRCh38
rs910310567 Chr.12: 67649774-67649774 on Build GRCh38
rs756266896 Chr.12: 67649908-67649908 on Build GRCh38
rs201364445 Chr.12: 67649771-67649771 on Build GRCh38
rs769222761 Chr.12: 67649870-67649870 on Build GRCh38
rs755620501 Chr.12: 67649971-67649979 on Build GRCh38
rs749747547 Chr.12: 67649830-67649830 on Build GRCh38
rs779382386 Chr.12: 67649846-67649846 on Build GRCh38
rs779248417 Chr.12: 67649944-67649944 on Build GRCh38
rs998584416 Chr.12: 67650005-67650005 on Build GRCh38
rs1030477020 Chr.12: 67650108-67650108 on Build GRCh38
rs778388363 Chr.12: 67649756-67649756 on Build GRCh38
rs752910064 Chr.12: 67649907-67649907 on Build GRCh38
rs188476944 Chr.12: 67650139-67650139 on Build GRCh38
rs562256886 Chr.12: 67650044-67650044 on Build GRCh38
rs778067609 Chr.12: 67649921-67649921 on Build GRCh38
rs951940891 Chr.12: 67650047-67650047 on Build GRCh38
rs539362642 Chr.12: 67649939-67649939 on Build GRCh38
rs915192868 Chr.12: 67649815-67649815 on Build GRCh38
rs112314435 Chr.12: 67649971-67649971 on Build GRCh38
rs531172685 Chr.12: 67650168-67650168 on Build GRCh38
rs1013035228 Chr.12: 67650006-67650006 on Build GRCh38
rs541376487 Chr.12: 67650084-67650084 on Build GRCh38
rs989537659 Chr.12: 67650045-67650045 on Build GRCh38
rs971362210 Chr.12: 67649736-67649736 on Build GRCh38
rs1046228890 Chr.12: 67650015-67650016 on Build GRCh38
rs766206981 Chr.12: 67649889-67649889 on Build GRCh38
rs973421867 Chr.12: 67649983-67649983 on Build GRCh38
rs945021919 Chr.12: 67649910-67649910 on Build GRCh38
rs756215589 Chr.12: 67649817-67649817 on Build GRCh38
rs776156501 Chr.12: 67649777-67649777 on Build GRCh38
rs993444347 Chr.12: 67650032-67650032 on Build GRCh38
rs976863732 Chr.12: 67649735-67649735 on Build GRCh38
rs575860144 Chr.12: 67649974-67649974 on Build GRCh38
rs901180065 Chr.12: 67649998-67649998 on Build GRCh38
rs966495589 Chr.12: 67650075-67650075 on Build GRCh38
rs894533257 Chr.12: 67649722-67649722 on Build GRCh38
rs750300664 Chr.12: 67649782-67649782 on Build GRCh38
rs557502236 Chr.12: 67649940-67649940 on Build GRCh38
rs763002690 Chr.12: 67649885-67649885 on Build GRCh38
rs755280655 Chr.12: 67649798-67649798 on Build GRCh38
rs767186702 Chr.12: 67649760-67649760 on Build GRCh38
rs761271046 Chr.12: 67649780-67649780 on Build GRCh38
rs535478382 Chr.12: 67649794-67649794 on Build GRCh38
rs574053939 Chr.12: 67650049-67650049 on Build GRCh38
rs781395476 Chr.12: 67649746-67649746 on Build GRCh38
rs1024273503 Chr.12: 67649732-67649732 on Build GRCh38
rs202146557 Chr.12: 67649781-67649781 on Build GRCh38
rs953944664 Chr.12: 67649779-67649779 on Build GRCh38
rs1013352856 Chr.12: 67649731-67649732 on Build GRCh38
rs1048530632 Chr.12: 67649970-67649970 on Build GRCh38
rs963358018 Chr.12: 67649977-67649977 on Build GRCh38
rs951908871 Chr.12: 67649764-67649764 on Build GRCh38
rs751412921 Chr.12: 67649890-67649890 on Build GRCh38
rs894884286 Chr.12: 67649954-67649954 on Build GRCh38
rs1006468768 Chr.12: 67650060-67650060 on Build GRCh38
rs779123015 Chr.12: 67650054-67650054 on Build GRCh38
rs1023716098 Chr.12: 67649987-67649987 on Build GRCh38
rs770467256 Chr.12: 67649750-67649750 on Build GRCh38
rs778439622 Chr.12: 67649825-67649825 on Build GRCh38
rs746265163 Chr.12: 67649967-67649967 on Build GRCh38
rs758629541 Chr.12: 67649973-67649973 on Build GRCh38
rs559803351 Chr.12: 67650095-67650095 on Build GRCh38
rs1028433392 Chr.12: 67650043-67650043 on Build GRCh38
rs964895495 Chr.12: 67650086-67650086 on Build GRCh38
rs762736260 Chr.12: 67649789-67649789 on Build GRCh38
rs752858847 Chr.12: 67649812-67649812 on Build GRCh38
rs916361111 Chr.12: 67650163-67650163 on Build GRCh38
rs948911301 Chr.12: 67649960-67649960 on Build GRCh38
rs1004438125 Chr.12: 67649981-67649981 on Build GRCh38
rs759216183 Chr.12: 67649892-67649892 on Build GRCh38
rs984560045 Chr.12: 67649765-67649765 on Build GRCh38
rs752701839 Chr.12: 67650159-67650162 on Build GRCh38
rs753955862 Chr.12: 67649923-67649923 on Build GRCh38
rs752199254 Chr.12: 67649929-67649940 on Build GRCh38
rs907234099 Chr.12: 67650029-67650029 on Build GRCh38
rs768385320 Chr.12: 67649852-67649852 on Build GRCh38
rs553490122 Chr.12: 67649803-67649803 on Build GRCh38
rs760443737 Chr.12: 67649772-67649772 on Build GRCh38
rs768171053 Chr.12: 67649776-67649776 on Build GRCh38
rs965733307 Chr.12: 67649734-67649734 on Build GRCh38
rs748220678 Chr.12: 67649748-67649748 on Build GRCh38
rs772702866 Chr.12: 67649874-67649874 on Build GRCh38
rs368559072 Chr.12: 67649906-67649906 on Build GRCh38
rs771339249 Chr.12: 67649840-67649840 on Build GRCh38
rs745890365 Chr.12: 67650071-67650071 on Build GRCh38
rs552133834 Chr.12: 67650132-67650132 on Build GRCh38
rs771272087 Chr.12: 67649759-67649759 on Build GRCh38
rs1017402044 Chr.12: 67650074-67650074 on Build GRCh38
rs183590629 Chr.12: 67649997-67649997 on Build GRCh38
rs939070981 Chr.12: 67650133-67650133 on Build GRCh38
rs936199571 Chr.12: 67650156-67650156 on Build GRCh38
rs567951314 Chr.12: 67649790-67649790 on Build GRCh38
rs924877323 Chr.12: 67650144-67650144 on Build GRCh38
rs533641810 Chr.12: 67650119-67650119 on Build GRCh38

View Full Product Details