Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67658192-67658697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
UniGene
Hs.173135
Gene Chromosome Location
Chr.12: 67648732-67665405 on Build GRCh38
Transcripts
NM_003583.3 NM_006482.2 XM_017020032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568592 c.1708T>G p.S570A Chr.12: 67658615-67658615 on Build GRCh38
2099014 c.1351C>T p.R451* Chr.12: 67658258-67658258 on Build GRCh38
5188140 c.1497G>A p.A499A Chr.12: 67658404-67658404 on Build GRCh38
4643758 c.1382A>G p.Y461C Chr.12: 67658289-67658289 on Build GRCh38
5622750 c.1624C>A p.P542T Chr.12: 67658531-67658531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769926495 Chr.12: 67658492-67658492 on Build GRCh38
rs745870753 Chr.12: 67658456-67658456 on Build GRCh38
rs749972090 Chr.12: 67658649-67658649 on Build GRCh38
rs575700478 Chr.12: 67658550-67658550 on Build GRCh38
rs746775763 Chr.12: 67658476-67658476 on Build GRCh38
rs760852696 Chr.12: 67658399-67658399 on Build GRCh38
rs201919838 Chr.12: 67658650-67658650 on Build GRCh38
rs188321581 Chr.12: 67658520-67658520 on Build GRCh38
rs146776716 Chr.12: 67658539-67658539 on Build GRCh38
rs753467166 Chr.12: 67658540-67658540 on Build GRCh38
rs919160707 Chr.12: 67658426-67658426 on Build GRCh38
rs755779810 Chr.12: 67658348-67658348 on Build GRCh38
rs3741644 Chr.12: 67658398-67658398 on Build GRCh38
rs749243922 Chr.12: 67658256-67658256 on Build GRCh38
rs200239121 Chr.12: 67658355-67658355 on Build GRCh38
rs766785374 Chr.12: 67658414-67658414 on Build GRCh38
rs748885684 Chr.12: 67658285-67658285 on Build GRCh38
rs535674843 Chr.12: 67658401-67658401 on Build GRCh38
rs749326516 Chr.12: 67658594-67658594 on Build GRCh38
rs369329175 Chr.12: 67658588-67658588 on Build GRCh38
rs1013767324 Chr.12: 67658560-67658560 on Build GRCh38
rs753315158 Chr.12: 67658445-67658445 on Build GRCh38
rs373302349 Chr.12: 67658609-67658609 on Build GRCh38
rs778950423 Chr.12: 67658365-67658365 on Build GRCh38
rs774611484 Chr.12: 67658519-67658519 on Build GRCh38
rs773264334 Chr.12: 67658499-67658499 on Build GRCh38
rs765747136 Chr.12: 67658402-67658402 on Build GRCh38
rs775800259 Chr.12: 67658293-67658293 on Build GRCh38
rs767994749 Chr.12: 67658610-67658610 on Build GRCh38
rs747669756 Chr.12: 67658226-67658226 on Build GRCh38
rs774866895 Chr.12: 67658607-67658607 on Build GRCh38
rs936839760 Chr.12: 67658504-67658504 on Build GRCh38
rs764940972 Chr.12: 67658636-67658636 on Build GRCh38
rs1028929571 Chr.12: 67658243-67658243 on Build GRCh38
rs573469138 Chr.12: 67658295-67658295 on Build GRCh38
rs755763829 Chr.12: 67658437-67658437 on Build GRCh38
rs375768980 Chr.12: 67658482-67658482 on Build GRCh38
rs189256234 Chr.12: 67658233-67658233 on Build GRCh38
rs765688297 Chr.12: 67658322-67658322 on Build GRCh38
rs943106175 Chr.12: 67658268-67658268 on Build GRCh38
rs780031437 Chr.12: 67658475-67658475 on Build GRCh38
rs954391793 Chr.12: 67658428-67658428 on Build GRCh38
rs375389205 Chr.12: 67658341-67658341 on Build GRCh38
rs780843951 Chr.12: 67658578-67658578 on Build GRCh38
rs772776286 Chr.12: 67658236-67658236 on Build GRCh38
rs35688869 Chr.12: 67658259-67658259 on Build GRCh38
rs777442529 Chr.12: 67658440-67658440 on Build GRCh38
rs771451192 Chr.12: 67658596-67658596 on Build GRCh38
rs766231212 Chr.12: 67658668-67658668 on Build GRCh38
rs777199119 Chr.12: 67658349-67658349 on Build GRCh38
rs748801595 Chr.12: 67658354-67658354 on Build GRCh38
rs754735753 Chr.12: 67658577-67658577 on Build GRCh38
rs749919220 Chr.12: 67658551-67658551 on Build GRCh38
rs577987133 Chr.12: 67658567-67658567 on Build GRCh38
rs769979169 Chr.12: 67658580-67658580 on Build GRCh38
rs77455066 Chr.12: 67658363-67658363 on Build GRCh38
rs748460025 Chr.12: 67658579-67658579 on Build GRCh38
rs751932333 Chr.12: 67658417-67658417 on Build GRCh38
rs201261546 Chr.12: 67658311-67658311 on Build GRCh38
rs369713542 Chr.12: 67658380-67658380 on Build GRCh38
rs56178357 Chr.12: 67658386-67658386 on Build GRCh38
rs138614034 Chr.12: 67658403-67658403 on Build GRCh38
rs867420742 Chr.12: 67658498-67658498 on Build GRCh38
rs779989013 Chr.12: 67658557-67658557 on Build GRCh38
rs138264226 Chr.12: 67658615-67658615 on Build GRCh38
rs770879931 Chr.12: 67658263-67658263 on Build GRCh38
rs61733656 Chr.12: 67658304-67658304 on Build GRCh38
rs910196972 Chr.12: 67658490-67658490 on Build GRCh38
rs768889751 Chr.12: 67658400-67658400 on Build GRCh38
rs954680330 Chr.12: 67658269-67658269 on Build GRCh38
rs777428293 Chr.12: 67658246-67658246 on Build GRCh38
rs772110790 Chr.12: 67658470-67658470 on Build GRCh38
rs143848143 Chr.12: 67658629-67658629 on Build GRCh38
rs756738175 Chr.12: 67658446-67658446 on Build GRCh38
rs774328160 Chr.12: 67658273-67658273 on Build GRCh38
rs989198070 Chr.12: 67658395-67658395 on Build GRCh38
rs758437734 Chr.12: 67658556-67658556 on Build GRCh38
rs776854870 Chr.12: 67658491-67658491 on Build GRCh38
rs147432605 Chr.12: 67658500-67658500 on Build GRCh38
rs55774594 Chr.12: 67658271-67658271 on Build GRCh38
rs772449209 Chr.12: 67658288-67658288 on Build GRCh38
rs769117431 Chr.12: 67658230-67658230 on Build GRCh38
rs535142121 Chr.12: 67658448-67658448 on Build GRCh38
rs763444454 Chr.12: 67658413-67658413 on Build GRCh38
rs372370232 Chr.12: 67658245-67658245 on Build GRCh38
rs758703731 Chr.12: 67658331-67658331 on Build GRCh38
rs750808823 Chr.12: 67658328-67658328 on Build GRCh38
rs780826338 Chr.12: 67658334-67658334 on Build GRCh38
rs544724491 Chr.12: 67658616-67658616 on Build GRCh38
rs759368728 Chr.12: 67658282-67658282 on Build GRCh38
rs772236701 Chr.12: 67658531-67658531 on Build GRCh38

View Full Product Details