Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 67658951-67659457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
-
UniGene
Hs.173135
Gene Chromosome Location
Chr.12: 67648732-67665405 on Build GRCh38
Transcripts
NM_003583.3 NM_006482.2 XM_017020032.1

SNP Detail

SNP ID Chromosome Location
rs747203503 Chr.12: 67659000-67659000 on Build GRCh38
rs549168836 Chr.12: 67659032-67659032 on Build GRCh38
rs138903572 Chr.12: 67659403-67659403 on Build GRCh38
rs1008713259 Chr.12: 67659376-67659376 on Build GRCh38
rs899262820 Chr.12: 67659027-67659027 on Build GRCh38
rs554721640 Chr.12: 67659238-67659238 on Build GRCh38
rs926880256 Chr.12: 67659204-67659204 on Build GRCh38
rs1010764302 Chr.12: 67659333-67659333 on Build GRCh38
rs940856655 Chr.12: 67659090-67659090 on Build GRCh38
rs955792424 Chr.12: 67659128-67659128 on Build GRCh38
rs547158894 Chr.12: 67659080-67659080 on Build GRCh38
rs573141502 Chr.12: 67659275-67659275 on Build GRCh38
rs1037903560 Chr.12: 67659126-67659126 on Build GRCh38
rs778779054 Chr.12: 67659350-67659350 on Build GRCh38
rs1047836742 Chr.12: 67659362-67659362 on Build GRCh38
rs546993706 Chr.12: 67658985-67658985 on Build GRCh38
rs900434559 Chr.12: 67659400-67659400 on Build GRCh38
rs559421504 Chr.12: 67658991-67658991 on Build GRCh38
rs1037728005 Chr.12: 67659271-67659271 on Build GRCh38
rs960403660 Chr.12: 67659404-67659404 on Build GRCh38
rs914104448 Chr.12: 67659160-67659160 on Build GRCh38
rs892349771 Chr.12: 67659277-67659277 on Build GRCh38
rs538611309 Chr.12: 67659159-67659159 on Build GRCh38
rs949550923 Chr.12: 67659181-67659181 on Build GRCh38
rs964756169 Chr.12: 67659113-67659113 on Build GRCh38
rs765357838 Chr.12: 67659374-67659374 on Build GRCh38
rs557335306 Chr.12: 67659177-67659177 on Build GRCh38
rs982793429 Chr.12: 67659199-67659199 on Build GRCh38
rs1033068050 Chr.12: 67659122-67659122 on Build GRCh38
rs1017519560 Chr.12: 67659081-67659081 on Build GRCh38
rs150233125 Chr.12: 67659379-67659379 on Build GRCh38
rs899315076 Chr.12: 67659154-67659154 on Build GRCh38
rs899127290 Chr.12: 67659301-67659301 on Build GRCh38
rs755207939 Chr.12: 67659427-67659427 on Build GRCh38
rs3741645 Chr.12: 67659037-67659037 on Build GRCh38
rs544641881 Chr.12: 67659396-67659396 on Build GRCh38
rs938006602 Chr.12: 67659004-67659004 on Build GRCh38
rs962314341 Chr.12: 67659426-67659426 on Build GRCh38
rs35797897 Chr.12: 67659092-67659093 on Build GRCh38
rs978464485 Chr.12: 67659119-67659119 on Build GRCh38
rs528400776 Chr.12: 67659266-67659266 on Build GRCh38
rs1038222128 Chr.12: 67659020-67659020 on Build GRCh38
rs757128866 Chr.12: 67659117-67659117 on Build GRCh38
rs151129263 Chr.12: 67659206-67659206 on Build GRCh38
rs905229492 Chr.12: 67659003-67659003 on Build GRCh38
rs535103840 Chr.12: 67659057-67659057 on Build GRCh38
rs1026156122 Chr.12: 67659040-67659040 on Build GRCh38
rs571613875 Chr.12: 67659097-67659097 on Build GRCh38
rs4913352 Chr.12: 67659371-67659371 on Build GRCh38
rs762014657 Chr.12: 67659295-67659295 on Build GRCh38
rs770926300 Chr.12: 67659205-67659205 on Build GRCh38
rs1032646510 Chr.12: 67659407-67659407 on Build GRCh38
rs577676225 Chr.12: 67659395-67659395 on Build GRCh38
rs540723911 Chr.12: 67659325-67659325 on Build GRCh38
rs753777047 Chr.12: 67659052-67659052 on Build GRCh38
rs530847692 Chr.12: 67659019-67659019 on Build GRCh38
rs141039166 Chr.12: 67659229-67659229 on Build GRCh38
rs955697122 Chr.12: 67659415-67659415 on Build GRCh38

View Full Product Details