Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 59774028-59774549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH3
Entrez Gene ID
81624
Gene Name
diaphanous related formin 3
Gene Aliases
AN, AUNA1, DIA2, DRF3, NSDAN, diap3, mDia2
UniGene
Hs.283127
Gene Chromosome Location
Chr.13: 59665583-60163985 on Build GRCh38
Transcripts
NM_001042517.1 NM_001258366.1 NM_001258367.1 NM_001258368.1 NM_001258369.1 NM_030932.3 XM_006719876.1 XM_011535258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355263 c.3319+9C>A p.? Chr.13: 59774180-59774180 on Build GRCh38
416330 c.3270G>A p.Q1090Q Chr.13: 59774238-59774238 on Build GRCh38
1180802 c.3260-6delT p.? Chr.13: 59774254-59774254 on Build GRCh38
4409190 c.3260A>G p.D1087G Chr.13: 59774248-59774248 on Build GRCh38
7135678 c.3292A>T p.R1098W Chr.13: 59774216-59774216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199814282 Chr.13: 59774225-59774225 on Build GRCh38
rs546373579 Chr.13: 59774062-59774062 on Build GRCh38
rs759456145 Chr.13: 59774242-59774242 on Build GRCh38
rs566429047 Chr.13: 59774351-59774351 on Build GRCh38
rs556066898 Chr.13: 59774260-59774260 on Build GRCh38
rs1047292000 Chr.13: 59774473-59774473 on Build GRCh38
rs1023039388 Chr.13: 59774122-59774122 on Build GRCh38
rs913311740 Chr.13: 59774319-59774322 on Build GRCh38
rs778077172 Chr.13: 59774230-59774230 on Build GRCh38
rs1000507082 Chr.13: 59774364-59774364 on Build GRCh38
rs201162217 Chr.13: 59774241-59774241 on Build GRCh38
rs1032206293 Chr.13: 59774369-59774369 on Build GRCh38
rs190669571 Chr.13: 59774178-59774178 on Build GRCh38
rs548120102 Chr.13: 59774348-59774348 on Build GRCh38
rs959215924 Chr.13: 59774068-59774068 on Build GRCh38
rs773219443 Chr.13: 59774265-59774265 on Build GRCh38
rs751261646 Chr.13: 59774261-59774261 on Build GRCh38
rs567082956 Chr.13: 59774215-59774215 on Build GRCh38
rs991088014 Chr.13: 59774113-59774113 on Build GRCh38
rs1039885178 Chr.13: 59774415-59774415 on Build GRCh38
rs766712682 Chr.13: 59774280-59774280 on Build GRCh38
rs564474219 Chr.13: 59774130-59774130 on Build GRCh38
rs750697558 Chr.13: 59774161-59774161 on Build GRCh38
rs774915340 Chr.13: 59774251-59774251 on Build GRCh38
rs535758021 Chr.13: 59774254-59774254 on Build GRCh38
rs774643709 Chr.13: 59774131-59774131 on Build GRCh38
rs1032235780 Chr.13: 59774057-59774057 on Build GRCh38
rs764704857 Chr.13: 59774142-59774142 on Build GRCh38
rs990598876 Chr.13: 59774281-59774281 on Build GRCh38
rs182505430 Chr.13: 59774146-59774146 on Build GRCh38
rs762540302 Chr.13: 59774259-59774259 on Build GRCh38
rs369779991 Chr.13: 59774238-59774238 on Build GRCh38
rs1013286655 Chr.13: 59774502-59774502 on Build GRCh38
rs943748980 Chr.13: 59774271-59774271 on Build GRCh38
rs942931567 Chr.13: 59774413-59774413 on Build GRCh38
rs755596264 Chr.13: 59774189-59774189 on Build GRCh38
rs1025188544 Chr.13: 59774456-59774456 on Build GRCh38
rs772310796 Chr.13: 59774251-59774252 on Build GRCh38
rs762590878 Chr.13: 59774153-59774153 on Build GRCh38
rs779486122 Chr.13: 59774499-59774499 on Build GRCh38
rs931080444 Chr.13: 59774446-59774446 on Build GRCh38
rs762566469 Chr.13: 59774267-59774270 on Build GRCh38
rs182878494 Chr.13: 59774430-59774430 on Build GRCh38
rs761654834 Chr.13: 59774253-59774254 on Build GRCh38
rs533791725 Chr.13: 59774400-59774400 on Build GRCh38
rs765298623 Chr.13: 59774254-59774255 on Build GRCh38
rs113570721 Chr.13: 59774133-59774133 on Build GRCh38
rs372237022 Chr.13: 59774179-59774179 on Build GRCh38
rs1013255520 Chr.13: 59774236-59774236 on Build GRCh38
rs186208845 Chr.13: 59774154-59774154 on Build GRCh38
rs886849811 Chr.13: 59774485-59774488 on Build GRCh38
rs547189359 Chr.13: 59774190-59774190 on Build GRCh38
rs78933251 Chr.13: 59774324-59774324 on Build GRCh38
rs745447228 Chr.13: 59774266-59774266 on Build GRCh38
rs746147763 Chr.13: 59774198-59774199 on Build GRCh38
rs761398957 Chr.13: 59774265-59774265 on Build GRCh38
rs780930028 Chr.13: 59774188-59774188 on Build GRCh38
rs558728989 Chr.13: 59774401-59774401 on Build GRCh38
rs61956702 Chr.13: 59774274-59774274 on Build GRCh38
rs772125942 Chr.13: 59774229-59774229 on Build GRCh38

View Full Product Details