Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 94461677-94462181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
UniGene
Hs.301865
Gene Chromosome Location
Chr.13: 94437304-94550265 on Build GRCh38
Transcripts
NM_001129889.2 NM_001322182.1 NM_001322183.1 NM_001322184.1 NM_001322185.1 NM_001322186.1 NM_001922.4 XM_011521049.1 XM_017020401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641833 c.876C>A p.Y292* Chr.13: 94462177-94462177 on Build GRCh38
6537996 c.951G>C p.M317I Chr.13: 94462102-94462102 on Build GRCh38
1629187 c.1043+5G>C p.? Chr.13: 94462005-94462005 on Build GRCh38
1629188 c.1043+5G>C p.? Chr.13: 94462005-94462005 on Build GRCh38
5431684 c.889A>G p.T297A Chr.13: 94462164-94462164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538118052 Chr.13: 94461889-94461889 on Build GRCh38
rs1033131827 Chr.13: 94461790-94461790 on Build GRCh38
rs1012184615 Chr.13: 94461733-94461733 on Build GRCh38
rs546808393 Chr.13: 94461902-94461902 on Build GRCh38
rs896120664 Chr.13: 94461720-94461720 on Build GRCh38
rs776517934 Chr.13: 94462084-94462084 on Build GRCh38
rs757604658 Chr.13: 94462051-94462051 on Build GRCh38
rs745762196 Chr.13: 94461843-94461843 on Build GRCh38
rs781489627 Chr.13: 94462069-94462069 on Build GRCh38
rs769559628 Chr.13: 94461980-94461980 on Build GRCh38
rs375498582 Chr.13: 94462103-94462103 on Build GRCh38
rs567029460 Chr.13: 94461944-94461944 on Build GRCh38
rs142117845 Chr.13: 94462068-94462068 on Build GRCh38
rs1014838267 Chr.13: 94461838-94461838 on Build GRCh38
rs761519974 Chr.13: 94461991-94461991 on Build GRCh38
rs368199467 Chr.13: 94462002-94462002 on Build GRCh38
rs979661507 Chr.13: 94461764-94461764 on Build GRCh38
rs151042383 Chr.13: 94461978-94461978 on Build GRCh38
rs778060068 Chr.13: 94462049-94462049 on Build GRCh38
rs768747820 Chr.13: 94462081-94462081 on Build GRCh38
rs907823788 Chr.13: 94461994-94461994 on Build GRCh38
rs892043679 Chr.13: 94461819-94461819 on Build GRCh38
rs780998962 Chr.13: 94461719-94461719 on Build GRCh38
rs746133761 Chr.13: 94462071-94462071 on Build GRCh38
rs771162877 Chr.13: 94461961-94461961 on Build GRCh38
rs970140852 Chr.13: 94461939-94461939 on Build GRCh38
rs367891461 Chr.13: 94462136-94462136 on Build GRCh38
rs536731053 Chr.13: 94461864-94461864 on Build GRCh38
rs201520554 Chr.13: 94462011-94462011 on Build GRCh38
rs148575940 Chr.13: 94462077-94462077 on Build GRCh38
rs760223077 Chr.13: 94462010-94462010 on Build GRCh38
rs897290246 Chr.13: 94461995-94461995 on Build GRCh38
rs142861036 Chr.13: 94461810-94461810 on Build GRCh38
rs1034687961 Chr.13: 94461706-94461706 on Build GRCh38
rs760281161 Chr.13: 94462127-94462127 on Build GRCh38
rs941573455 Chr.13: 94461907-94461907 on Build GRCh38
rs74325354 Chr.13: 94462144-94462144 on Build GRCh38
rs911453130 Chr.13: 94461860-94461860 on Build GRCh38
rs140917943 Chr.13: 94462076-94462076 on Build GRCh38
rs758950858 Chr.13: 94462048-94462048 on Build GRCh38
rs753213570 Chr.13: 94462046-94462046 on Build GRCh38
rs774311825 Chr.13: 94461989-94461989 on Build GRCh38
rs201741628 Chr.13: 94461986-94461986 on Build GRCh38
rs985757028 Chr.13: 94461859-94461859 on Build GRCh38
rs1011900553 Chr.13: 94462066-94462066 on Build GRCh38
rs768551461 Chr.13: 94461987-94461987 on Build GRCh38
rs781091100 Chr.13: 94461974-94461974 on Build GRCh38
rs763407563 Chr.13: 94462145-94462145 on Build GRCh38
rs756039485 Chr.13: 94462108-94462108 on Build GRCh38
rs967931190 Chr.13: 94461755-94461755 on Build GRCh38
rs999185265 Chr.13: 94461788-94461788 on Build GRCh38
rs772918072 Chr.13: 94462120-94462120 on Build GRCh38
rs1043386900 Chr.13: 94461734-94461734 on Build GRCh38
rs764988394 Chr.13: 94461882-94461882 on Build GRCh38
rs767286434 Chr.13: 94461997-94461997 on Build GRCh38
rs1027722208 Chr.13: 94461960-94461960 on Build GRCh38
rs1023950240 Chr.13: 94461744-94461744 on Build GRCh38
rs747973265 Chr.13: 94462101-94462101 on Build GRCh38
rs141684345 Chr.13: 94461726-94461726 on Build GRCh38
rs751950305 Chr.13: 94462067-94462067 on Build GRCh38
rs922611824 Chr.13: 94461835-94461835 on Build GRCh38
rs568976756 Chr.13: 94461736-94461736 on Build GRCh38
rs956502652 Chr.13: 94462157-94462157 on Build GRCh38
rs1023580929 Chr.13: 94462070-94462070 on Build GRCh38
rs1024440418 Chr.13: 94461880-94461880 on Build GRCh38
rs774224411 Chr.13: 94462095-94462095 on Build GRCh38
rs988815536 Chr.13: 94461832-94461832 on Build GRCh38
rs749724235 Chr.13: 94462019-94462019 on Build GRCh38
rs983463036 Chr.13: 94461981-94461981 on Build GRCh38
rs935027499 Chr.13: 94462135-94462135 on Build GRCh38

View Full Product Details