Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24000517-24001020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS4L2
Entrez Gene ID
317749
Gene Name
dehydrogenase/reductase 4 like 2
Gene Aliases
SDR25C3
UniGene
Hs.743442
Gene Chromosome Location
Chr.14: 23969874-24006408 on Build GRCh38
Transcripts
NM_001193635.1 NM_001193636.1 NM_001193637.1 NM_198083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
698127 c.476G>A p.R159Q Chr.14: 24000930-24000930 on Build GRCh38
5784072 c.473+8G>A p.? Chr.14: 24000941-24000941 on Build GRCh38
5784071 c.479+8G>A p.? Chr.14: 24000941-24000941 on Build GRCh38
4419847 c.475C>T p.R159* Chr.14: 24000929-24000929 on Build GRCh38
4419848 c.469C>T p.R157* Chr.14: 24000929-24000929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376969886 Chr.14: 24000941-24000941 on Build GRCh38
rs557269720 Chr.14: 24000921-24000921 on Build GRCh38
rs757474367 Chr.14: 24000954-24000954 on Build GRCh38
rs781132993 Chr.14: 24000965-24000965 on Build GRCh38
rs772953007 Chr.14: 24000905-24000905 on Build GRCh38
rs61334310 Chr.14: 24000990-24000990 on Build GRCh38
rs769603521 Chr.14: 24000966-24000966 on Build GRCh38
rs376746348 Chr.14: 24000854-24000854 on Build GRCh38
rs772930070 Chr.14: 24000969-24000969 on Build GRCh38
rs768414851 Chr.14: 24000825-24000825 on Build GRCh38
rs1045179275 Chr.14: 24000620-24000620 on Build GRCh38
rs887538068 Chr.14: 24000639-24000639 on Build GRCh38
rs772352191 Chr.14: 24000644-24000644 on Build GRCh38
rs753061153 Chr.14: 24000943-24000943 on Build GRCh38
rs775527434 Chr.14: 24000817-24000817 on Build GRCh38
rs563626521 Chr.14: 24000993-24000993 on Build GRCh38
rs997363998 Chr.14: 24000702-24000702 on Build GRCh38
rs564645090 Chr.14: 24000977-24000977 on Build GRCh38
rs748957064 Chr.14: 24000970-24000970 on Build GRCh38
rs1036087857 Chr.14: 24000976-24000976 on Build GRCh38
rs572457229 Chr.14: 24000959-24000959 on Build GRCh38
rs968822106 Chr.14: 24000797-24000797 on Build GRCh38
rs1014703297 Chr.14: 24000679-24000679 on Build GRCh38
rs201378287 Chr.14: 24000947-24000947 on Build GRCh38
rs961875592 Chr.14: 24000685-24000685 on Build GRCh38
rs528838051 Chr.14: 24000737-24000737 on Build GRCh38
rs1030217988 Chr.14: 24000715-24000715 on Build GRCh38
rs748936963 Chr.14: 24000844-24000846 on Build GRCh38
rs369323650 Chr.14: 24000864-24000864 on Build GRCh38
rs28535492 Chr.14: 24000971-24000971 on Build GRCh38
rs55700022 Chr.14: 24001000-24001000 on Build GRCh38
rs768644359 Chr.14: 24000898-24000898 on Build GRCh38
rs372826656 Chr.14: 24000930-24000930 on Build GRCh38
rs1011930549 Chr.14: 24000583-24000583 on Build GRCh38
rs369877396 Chr.14: 24000904-24000904 on Build GRCh38
rs754854961 Chr.14: 24000961-24000961 on Build GRCh38
rs749849127 Chr.14: 24000844-24000844 on Build GRCh38
rs200895612 Chr.14: 24000998-24000998 on Build GRCh38
rs914593329 Chr.14: 24000747-24000747 on Build GRCh38
rs571342578 Chr.14: 24000889-24000889 on Build GRCh38
rs866434008 Chr.14: 24000752-24000752 on Build GRCh38
rs375966037 Chr.14: 24000879-24000879 on Build GRCh38
rs761313175 Chr.14: 24000992-24000992 on Build GRCh38
rs778079091 Chr.14: 24000880-24000880 on Build GRCh38
rs761446403 Chr.14: 24000830-24000830 on Build GRCh38
rs1006423235 Chr.14: 24000641-24000641 on Build GRCh38
rs376879565 Chr.14: 24000995-24000995 on Build GRCh38
rs1057410823 Chr.14: 24000574-24000574 on Build GRCh38
rs76909974 Chr.14: 24000978-24000978 on Build GRCh38
rs375757538 Chr.14: 24000935-24000935 on Build GRCh38
rs59027712 Chr.14: 24000667-24000667 on Build GRCh38
rs565736401 Chr.14: 24000847-24000847 on Build GRCh38
rs896133568 Chr.14: 24000576-24000576 on Build GRCh38
rs144805644 Chr.14: 24000958-24000958 on Build GRCh38
rs986149035 Chr.14: 24000728-24000728 on Build GRCh38
rs570063294 Chr.14: 24000683-24000683 on Build GRCh38
rs953468291 Chr.14: 24000716-24000716 on Build GRCh38
rs537286969 Chr.14: 24000897-24000897 on Build GRCh38
rs370529426 Chr.14: 24000983-24000983 on Build GRCh38
rs372465757 Chr.14: 24000849-24000849 on Build GRCh38
rs1811890 Chr.14: 24000929-24000929 on Build GRCh38
rs754065784 Chr.14: 24000950-24000950 on Build GRCh38
rs764877248 Chr.14: 24000837-24000837 on Build GRCh38
rs776463189 Chr.14: 24000829-24000829 on Build GRCh38
rs776337533 Chr.14: 24000900-24000900 on Build GRCh38
rs756575609 Chr.14: 24000946-24000946 on Build GRCh38
rs770425355 Chr.14: 24000932-24000932 on Build GRCh38
rs368156723 Chr.14: 24000820-24000820 on Build GRCh38
rs371326305 Chr.14: 24000878-24000878 on Build GRCh38
rs755594079 Chr.14: 24000856-24000856 on Build GRCh38
rs771010677 Chr.14: 24000894-24000894 on Build GRCh38

View Full Product Details