Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 87933473-87933980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALC
Entrez Gene ID
2581
Gene Name
galactosylceramidase
Gene Aliases
-
UniGene
Hs.513439
Gene Chromosome Location
Chr.14: 87933014-87993665 on Build GRCh38
Transcripts
NM_000153.3 NM_001201401.1 NM_001201402.1 XM_011536618.2

SNP Detail

SNP ID Chromosome Location
rs975815066 Chr.14: 87933512-87933512 on Build GRCh38
rs370559 Chr.14: 87933743-87933743 on Build GRCh38
rs1005299395 Chr.14: 87933727-87933727 on Build GRCh38
rs772671366 Chr.14: 87933834-87933834 on Build GRCh38
rs149009218 Chr.14: 87933954-87933954 on Build GRCh38
rs894879931 Chr.14: 87933697-87933697 on Build GRCh38
rs1018308602 Chr.14: 87933818-87933818 on Build GRCh38
rs530684407 Chr.14: 87933907-87933907 on Build GRCh38
rs985442758 Chr.14: 87933806-87933806 on Build GRCh38
rs1045763576 Chr.14: 87933780-87933780 on Build GRCh38
rs1803611 Chr.14: 87933869-87933869 on Build GRCh38
rs749541522 Chr.14: 87933498-87933498 on Build GRCh38
rs375388813 Chr.14: 87933549-87933549 on Build GRCh38
rs893278017 Chr.14: 87933824-87933824 on Build GRCh38
rs548211859 Chr.14: 87933750-87933750 on Build GRCh38
rs945208326 Chr.14: 87933891-87933891 on Build GRCh38
rs1029890728 Chr.14: 87933775-87933775 on Build GRCh38
rs1003136252 Chr.14: 87933649-87933649 on Build GRCh38
rs997684608 Chr.14: 87933907-87933908 on Build GRCh38
rs867813345 Chr.14: 87933626-87933626 on Build GRCh38
rs141199615 Chr.14: 87933544-87933546 on Build GRCh38
rs114174103 Chr.14: 87933931-87933931 on Build GRCh38
rs990266186 Chr.14: 87933857-87933857 on Build GRCh38
rs1054748501 Chr.14: 87933676-87933676 on Build GRCh38
rs200156630 Chr.14: 87933546-87933546 on Build GRCh38
rs1042035 Chr.14: 87933606-87933606 on Build GRCh38
rs116351733 Chr.14: 87933749-87933749 on Build GRCh38
rs545714854 Chr.14: 87933823-87933823 on Build GRCh38
rs751205211 Chr.14: 87933638-87933638 on Build GRCh38
rs748637699 Chr.14: 87933784-87933784 on Build GRCh38
rs536915624 Chr.14: 87933734-87933734 on Build GRCh38
rs185181747 Chr.14: 87933855-87933855 on Build GRCh38
rs978345613 Chr.14: 87933902-87933902 on Build GRCh38
rs967189458 Chr.14: 87933889-87933889 on Build GRCh38
rs939295930 Chr.14: 87933911-87933911 on Build GRCh38
rs1042270201 Chr.14: 87933590-87933590 on Build GRCh38
rs955620835 Chr.14: 87933798-87933798 on Build GRCh38
rs909027786 Chr.14: 87933680-87933680 on Build GRCh38
rs1054717273 Chr.14: 87933929-87933929 on Build GRCh38
rs560375239 Chr.14: 87933828-87933828 on Build GRCh38
rs554100237 Chr.14: 87933535-87933535 on Build GRCh38
rs913311444 Chr.14: 87933862-87933862 on Build GRCh38
rs941881990 Chr.14: 87933694-87933694 on Build GRCh38
rs773169886 Chr.14: 87933543-87933544 on Build GRCh38
rs941039886 Chr.14: 87933950-87933950 on Build GRCh38
rs907203541 Chr.14: 87933817-87933817 on Build GRCh38

View Full Product Details