Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30928461-30928962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAN1
Entrez Gene ID
22909
Gene Name
FANCD2/FANCI-associated nuclease 1
Gene Aliases
KIAA1018, KMIN, MTMR15, hFAN1
UniGene
Hs.584863
Gene Chromosome Location
Chr.15: 30903695-30943108 on Build GRCh38
Transcripts
NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_017022012.1 XM_017022013.1 XM_011521737.2

Gene Symbol
MTMR10
Entrez Gene ID
54893
Gene Name
myotubularin related protein 10
Gene Aliases
-
UniGene
Hs.30141
Gene Chromosome Location
Chr.15: 30919764-30991604 on Build GRCh38
Transcripts
NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_017022012.1 XM_017022013.1 XM_011521737.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660389 c.2500G>A p.E834K Chr.15: 30928564-30928564 on Build GRCh38
6396223 c.2534T>C p.L845P Chr.15: 30928598-30928598 on Build GRCh38
6660382 c.2583C>T p.N861N Chr.15: 30928647-30928647 on Build GRCh38
6660392 c.2568G>A p.P856P Chr.15: 30928632-30928632 on Build GRCh38
6451352 c.2586C>A p.A862A Chr.15: 30928650-30928650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758207030 Chr.15: 30928697-30928697 on Build GRCh38
rs143970863 Chr.15: 30928506-30928507 on Build GRCh38
rs760483915 Chr.15: 30928512-30928513 on Build GRCh38
rs58652305 Chr.15: 30928507-30928508 on Build GRCh38
rs775365256 Chr.15: 30928510-30928511 on Build GRCh38
rs113688207 Chr.15: 30928929-30928929 on Build GRCh38
rs1026950082 Chr.15: 30928873-30928873 on Build GRCh38
rs764214947 Chr.15: 30928548-30928548 on Build GRCh38
rs763258710 Chr.15: 30928582-30928582 on Build GRCh38
rs776742586 Chr.15: 30928648-30928648 on Build GRCh38
rs768157749 Chr.15: 30928670-30928670 on Build GRCh38
rs745669441 Chr.15: 30928541-30928541 on Build GRCh38
rs1017810823 Chr.15: 30928885-30928885 on Build GRCh38
rs61136501 Chr.15: 30928540-30928541 on Build GRCh38
rs904946116 Chr.15: 30928724-30928724 on Build GRCh38
rs368345932 Chr.15: 30928662-30928662 on Build GRCh38
rs776276752 Chr.15: 30928544-30928545 on Build GRCh38
rs879010975 Chr.15: 30928507-30928510 on Build GRCh38
rs1009069677 Chr.15: 30928879-30928879 on Build GRCh38
rs964778270 Chr.15: 30928899-30928899 on Build GRCh38
rs577593166 Chr.15: 30928935-30928935 on Build GRCh38
rs955019263 Chr.15: 30928875-30928875 on Build GRCh38
rs748299790 Chr.15: 30928503-30928504 on Build GRCh38
rs767651793 Chr.15: 30928654-30928654 on Build GRCh38
rs763854539 Chr.15: 30928515-30928516 on Build GRCh38
rs1027724111 Chr.15: 30928800-30928800 on Build GRCh38
rs753566292 Chr.15: 30928524-30928525 on Build GRCh38
rs373899691 Chr.15: 30928506-30928506 on Build GRCh38
rs537551224 Chr.15: 30928728-30928728 on Build GRCh38
rs535122436 Chr.15: 30928547-30928547 on Build GRCh38
rs746985218 Chr.15: 30928641-30928641 on Build GRCh38
rs781684033 Chr.15: 30928534-30928535 on Build GRCh38
rs766518053 Chr.15: 30928818-30928818 on Build GRCh38
rs993597417 Chr.15: 30928508-30928508 on Build GRCh38
rs367829539 Chr.15: 30928564-30928564 on Build GRCh38
rs147488785 Chr.15: 30928626-30928626 on Build GRCh38
rs756918589 Chr.15: 30928619-30928619 on Build GRCh38
rs774771247 Chr.15: 30928630-30928630 on Build GRCh38
rs570220005 Chr.15: 30928835-30928835 on Build GRCh38
rs200756403 Chr.15: 30928647-30928647 on Build GRCh38
rs777019964 Chr.15: 30928504-30928505 on Build GRCh38
rs142786123 Chr.15: 30928590-30928590 on Build GRCh38
rs749916559 Chr.15: 30928526-30928527 on Build GRCh38
rs372779313 Chr.15: 30928712-30928712 on Build GRCh38
rs796825695 Chr.15: 30928504-30928505 on Build GRCh38
rs750100150 Chr.15: 30928695-30928695 on Build GRCh38
rs981359752 Chr.15: 30928887-30928887 on Build GRCh38
rs774334166 Chr.15: 30928525-30928525 on Build GRCh38
rs764802743 Chr.15: 30928603-30928604 on Build GRCh38
rs993182217 Chr.15: 30928807-30928807 on Build GRCh38
rs181692230 Chr.15: 30928925-30928925 on Build GRCh38
rs771776626 Chr.15: 30928631-30928631 on Build GRCh38
rs549982773 Chr.15: 30928797-30928797 on Build GRCh38
rs760166456 Chr.15: 30928669-30928669 on Build GRCh38
rs753355065 Chr.15: 30928673-30928673 on Build GRCh38
rs993957751 Chr.15: 30928798-30928798 on Build GRCh38
rs574727900 Chr.15: 30928586-30928586 on Build GRCh38
rs778569968 Chr.15: 30928680-30928680 on Build GRCh38
rs746945535 Chr.15: 30928703-30928703 on Build GRCh38
rs1037355462 Chr.15: 30928793-30928793 on Build GRCh38
rs547920265 Chr.15: 30928735-30928735 on Build GRCh38
rs552242762 Chr.15: 30928865-30928865 on Build GRCh38
rs769919620 Chr.15: 30928539-30928540 on Build GRCh38
rs537740534 Chr.15: 30928710-30928710 on Build GRCh38
rs779752276 Chr.15: 30928699-30928699 on Build GRCh38
rs1026587573 Chr.15: 30928632-30928632 on Build GRCh38
rs897842995 Chr.15: 30928796-30928796 on Build GRCh38
rs760574027 Chr.15: 30928541-30928544 on Build GRCh38
rs763322755 Chr.15: 30928660-30928660 on Build GRCh38
rs750488618 Chr.15: 30928684-30928684 on Build GRCh38
rs777510680 Chr.15: 30928520-30928520 on Build GRCh38
rs139319999 Chr.15: 30928633-30928633 on Build GRCh38
rs755691510 Chr.15: 30928675-30928675 on Build GRCh38
rs144447348 Chr.15: 30928930-30928930 on Build GRCh38
rs188512595 Chr.15: 30928838-30928838 on Build GRCh38
rs747474438 Chr.15: 30928529-30928530 on Build GRCh38
rs180765412 Chr.15: 30928560-30928560 on Build GRCh38
rs914311782 Chr.15: 30928934-30928934 on Build GRCh38
rs776828162 Chr.15: 30928558-30928558 on Build GRCh38
rs963643211 Chr.15: 30928497-30928497 on Build GRCh38
rs572409384 Chr.15: 30928492-30928492 on Build GRCh38
rs184117422 Chr.15: 30928767-30928767 on Build GRCh38
rs769964560 Chr.15: 30928651-30928651 on Build GRCh38
rs756828695 Chr.15: 30928679-30928679 on Build GRCh38
rs566113394 Chr.15: 30928918-30928918 on Build GRCh38
rs767443420 Chr.15: 30928509-30928510 on Build GRCh38
rs770816763 Chr.15: 30928523-30928523 on Build GRCh38
rs755447386 Chr.15: 30928530-30928531 on Build GRCh38
rs898796025 Chr.15: 30928801-30928801 on Build GRCh38
rs763029297 Chr.15: 30928505-30928506 on Build GRCh38
rs543726663 Chr.15: 30928589-30928589 on Build GRCh38
rs1015919835 Chr.15: 30928493-30928493 on Build GRCh38
rs1052757175 Chr.15: 30928687-30928687 on Build GRCh38
rs374193799 Chr.15: 30928563-30928563 on Build GRCh38
rs561129086 Chr.15: 30928751-30928751 on Build GRCh38
rs1016486118 Chr.15: 30928727-30928727 on Build GRCh38
rs780740952 Chr.15: 30928528-30928529 on Build GRCh38
rs775455336 Chr.15: 30928541-30928543 on Build GRCh38
rs777494363 Chr.15: 30928595-30928595 on Build GRCh38
rs749009487 Chr.15: 30928522-30928522 on Build GRCh38

View Full Product Details