Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38324533-38324954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4054216 c.422A>C p.Q141P Chr.15: 38324808-38324808 on Build GRCh38
6333174 c.409G>A p.A137T Chr.15: 38324795-38324795 on Build GRCh38
5040700 c.413A>C p.D138A Chr.15: 38324799-38324799 on Build GRCh38
5610686 c.383C>A p.P128H Chr.15: 38324769-38324769 on Build GRCh38
470547 c.395A>G p.N132S Chr.15: 38324781-38324781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139396722 Chr.15: 38324562-38324562 on Build GRCh38
rs755991015 Chr.15: 38324786-38324786 on Build GRCh38
rs551043433 Chr.15: 38324559-38324559 on Build GRCh38
rs772830013 Chr.15: 38324782-38324782 on Build GRCh38
rs368681912 Chr.15: 38324843-38324843 on Build GRCh38
rs767491326 Chr.15: 38324829-38324829 on Build GRCh38
rs967177024 Chr.15: 38324868-38324868 on Build GRCh38
rs765313507 Chr.15: 38324604-38324604 on Build GRCh38
rs375647156 Chr.15: 38324731-38324731 on Build GRCh38
rs12148911 Chr.15: 38324803-38324803 on Build GRCh38
rs1039898669 Chr.15: 38324601-38324601 on Build GRCh38
rs754866278 Chr.15: 38324749-38324749 on Build GRCh38
rs765800186 Chr.15: 38324794-38324794 on Build GRCh38
rs778868909 Chr.15: 38324791-38324791 on Build GRCh38
rs548225952 Chr.15: 38324593-38324593 on Build GRCh38
rs751138842 Chr.15: 38324799-38324799 on Build GRCh38
rs754814266 Chr.15: 38324824-38324824 on Build GRCh38
rs939893605 Chr.15: 38324664-38324664 on Build GRCh38
rs1879937 Chr.15: 38324914-38324914 on Build GRCh38
rs1026261584 Chr.15: 38324845-38324845 on Build GRCh38
rs758934848 Chr.15: 38324736-38324739 on Build GRCh38
rs992525037 Chr.15: 38324611-38324611 on Build GRCh38
rs749329100 Chr.15: 38324729-38324729 on Build GRCh38
rs764823722 Chr.15: 38324771-38324771 on Build GRCh38
rs761058615 Chr.15: 38324770-38324770 on Build GRCh38
rs12595839 Chr.15: 38324861-38324861 on Build GRCh38
rs1037259379 Chr.15: 38324685-38324685 on Build GRCh38
rs149883080 Chr.15: 38324774-38324774 on Build GRCh38
rs757559269 Chr.15: 38324850-38324850 on Build GRCh38
rs778167838 Chr.15: 38324726-38324726 on Build GRCh38
rs200319824 Chr.15: 38324834-38324834 on Build GRCh38
rs762633609 Chr.15: 38324787-38324787 on Build GRCh38
rs575553986 Chr.15: 38324897-38324897 on Build GRCh38
rs748694682 Chr.15: 38324717-38324717 on Build GRCh38
rs960986516 Chr.15: 38324916-38324916 on Build GRCh38
rs371837252 Chr.15: 38324751-38324751 on Build GRCh38
rs113547130 Chr.15: 38324849-38324849 on Build GRCh38
rs376134678 Chr.15: 38324753-38324753 on Build GRCh38
rs909981334 Chr.15: 38324585-38324585 on Build GRCh38
rs367973545 Chr.15: 38324734-38324734 on Build GRCh38
rs1879939 Chr.15: 38324614-38324614 on Build GRCh38
rs1010583227 Chr.15: 38324691-38324691 on Build GRCh38
rs934232696 Chr.15: 38324612-38324612 on Build GRCh38
rs922799267 Chr.15: 38324604-38324604 on Build GRCh38
rs184294214 Chr.15: 38324603-38324603 on Build GRCh38
rs984599734 Chr.15: 38324582-38324582 on Build GRCh38
rs960932830 Chr.15: 38324604-38324605 on Build GRCh38
rs1048108790 Chr.15: 38324625-38324625 on Build GRCh38
rs770391922 Chr.15: 38324741-38324744 on Build GRCh38
rs755992043 Chr.15: 38324847-38324847 on Build GRCh38
rs774560090 Chr.15: 38324733-38324733 on Build GRCh38
rs386783166 Chr.15: 38324912-38324914 on Build GRCh38
rs778804596 Chr.15: 38324855-38324855 on Build GRCh38
rs1879938 Chr.15: 38324912-38324912 on Build GRCh38
rs190367864 Chr.15: 38324642-38324642 on Build GRCh38

View Full Product Details