Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22258374-22258871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2K
Entrez Gene ID
29904
Gene Name
eukaryotic elongation factor 2 kinase
Gene Aliases
HSU93850, eEF-2K
UniGene
Hs.498892
Gene Chromosome Location
Chr.16: 22206264-22288745 on Build GRCh38
Transcripts
NM_013302.3 XM_017023193.1 XM_017023194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376693 c.1036G>T p.A346S Chr.16: 22258500-22258500 on Build GRCh38
6522660 c.1149G>A p.E383E Chr.16: 22258613-22258613 on Build GRCh38
1376695 c.1161C>T p.D387D Chr.16: 22258625-22258625 on Build GRCh38
393273 c.1149G>T p.E383D Chr.16: 22258613-22258613 on Build GRCh38
5603454 c.1179C>T p.L393L Chr.16: 22258643-22258643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879097209 Chr.16: 22258438-22258438 on Build GRCh38
rs529060079 Chr.16: 22258661-22258661 on Build GRCh38
rs950984411 Chr.16: 22258552-22258552 on Build GRCh38
rs761579720 Chr.16: 22258589-22258589 on Build GRCh38
rs200544061 Chr.16: 22258695-22258695 on Build GRCh38
rs144592275 Chr.16: 22258676-22258676 on Build GRCh38
rs573803833 Chr.16: 22258816-22258816 on Build GRCh38
rs759547277 Chr.16: 22258616-22258616 on Build GRCh38
rs763605770 Chr.16: 22258807-22258807 on Build GRCh38
rs200714248 Chr.16: 22258500-22258500 on Build GRCh38
rs183300451 Chr.16: 22258584-22258584 on Build GRCh38
rs757889253 Chr.16: 22258555-22258555 on Build GRCh38
rs201198682 Chr.16: 22258533-22258533 on Build GRCh38
rs368839566 Chr.16: 22258545-22258545 on Build GRCh38
rs971409660 Chr.16: 22258796-22258796 on Build GRCh38
rs941512529 Chr.16: 22258700-22258700 on Build GRCh38
rs775206083 Chr.16: 22258496-22258496 on Build GRCh38
rs957487224 Chr.16: 22258815-22258815 on Build GRCh38
rs767583545 Chr.16: 22258619-22258619 on Build GRCh38
rs144330987 Chr.16: 22258462-22258462 on Build GRCh38
rs751134098 Chr.16: 22258735-22258735 on Build GRCh38
rs148528298 Chr.16: 22258436-22258436 on Build GRCh38
rs946616863 Chr.16: 22258420-22258420 on Build GRCh38
rs754679239 Chr.16: 22258667-22258667 on Build GRCh38
rs775763205 Chr.16: 22258702-22258702 on Build GRCh38
rs745362197 Chr.16: 22258490-22258490 on Build GRCh38
rs1001856850 Chr.16: 22258708-22258708 on Build GRCh38
rs746496365 Chr.16: 22258666-22258666 on Build GRCh38
rs1017107138 Chr.16: 22258567-22258567 on Build GRCh38
rs187968648 Chr.16: 22258610-22258610 on Build GRCh38
rs200364752 Chr.16: 22258709-22258709 on Build GRCh38
rs148711774 Chr.16: 22258634-22258634 on Build GRCh38
rs748841544 Chr.16: 22258676-22258676 on Build GRCh38
rs751644939 Chr.16: 22258523-22258523 on Build GRCh38
rs745439125 Chr.16: 22258569-22258569 on Build GRCh38
rs754415093 Chr.16: 22258750-22258750 on Build GRCh38
rs1024503460 Chr.16: 22258772-22258772 on Build GRCh38
rs4783453 Chr.16: 22258546-22258546 on Build GRCh38
rs769572609 Chr.16: 22258677-22258677 on Build GRCh38
rs377220765 Chr.16: 22258606-22258606 on Build GRCh38
rs201712494 Chr.16: 22258660-22258660 on Build GRCh38
rs142158605 Chr.16: 22258498-22258498 on Build GRCh38
rs144252213 Chr.16: 22258622-22258622 on Build GRCh38
rs1009731450 Chr.16: 22258825-22258825 on Build GRCh38
rs778437391 Chr.16: 22258463-22258463 on Build GRCh38
rs1047103024 Chr.16: 22258694-22258694 on Build GRCh38
rs370205478 Chr.16: 22258716-22258716 on Build GRCh38
rs746798941 Chr.16: 22258710-22258710 on Build GRCh38
rs115217797 Chr.16: 22258705-22258705 on Build GRCh38
rs771682445 Chr.16: 22258493-22258493 on Build GRCh38
rs374751930 Chr.16: 22258522-22258522 on Build GRCh38
rs780182579 Chr.16: 22258625-22258625 on Build GRCh38
rs749763725 Chr.16: 22258478-22258478 on Build GRCh38
rs777547270 Chr.16: 22258557-22258558 on Build GRCh38
rs201576066 Chr.16: 22258570-22258570 on Build GRCh38
rs372222067 Chr.16: 22258509-22258509 on Build GRCh38
rs375505959 Chr.16: 22258459-22258459 on Build GRCh38
rs773044439 Chr.16: 22258688-22258688 on Build GRCh38
rs746619297 Chr.16: 22258577-22258577 on Build GRCh38
rs934198895 Chr.16: 22258481-22258481 on Build GRCh38
rs538124442 Chr.16: 22258696-22258696 on Build GRCh38
rs975392237 Chr.16: 22258674-22258674 on Build GRCh38
rs779295321 Chr.16: 22258635-22258635 on Build GRCh38
rs1043099277 Chr.16: 22258429-22258429 on Build GRCh38
rs781460960 Chr.16: 22258448-22258448 on Build GRCh38
rs980053968 Chr.16: 22258797-22258797 on Build GRCh38
rs756569293 Chr.16: 22258542-22258542 on Build GRCh38
rs752905138 Chr.16: 22258534-22258534 on Build GRCh38
rs146119676 Chr.16: 22258484-22258484 on Build GRCh38
rs202165153 Chr.16: 22258727-22258727 on Build GRCh38
rs750887173 Chr.16: 22258655-22258655 on Build GRCh38
rs868306819 Chr.16: 22258583-22258583 on Build GRCh38
rs756589486 Chr.16: 22258488-22258488 on Build GRCh38
rs934067489 Chr.16: 22258692-22258692 on Build GRCh38
rs770646648 Chr.16: 22258706-22258707 on Build GRCh38
rs749489869 Chr.16: 22258626-22258626 on Build GRCh38
rs758988450 Chr.16: 22258717-22258717 on Build GRCh38
rs761018908 Chr.16: 22258623-22258623 on Build GRCh38
rs770596342 Chr.16: 22258479-22258479 on Build GRCh38
rs779463321 Chr.16: 22258558-22258558 on Build GRCh38
rs754314099 Chr.16: 22258631-22258631 on Build GRCh38
rs138328974 Chr.16: 22258571-22258571 on Build GRCh38
rs368245066 Chr.16: 22258704-22258704 on Build GRCh38
rs149639518 Chr.16: 22258585-22258585 on Build GRCh38
rs890768684 Chr.16: 22258455-22258455 on Build GRCh38
rs200099793 Chr.16: 22258508-22258508 on Build GRCh38

View Full Product Details