Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30667461-30667958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBRS
Entrez Gene ID
64319
Gene Name
fibrosin
Gene Aliases
FBS, FBS1
UniGene
Hs.247186
Gene Chromosome Location
Chr.16: 30658142-30670810 on Build GRCh38
Transcripts
NM_001105079.2 XM_011545916.2 XM_011545917.2 XM_011545919.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3817815 c.220G>T p.G74* Chr.16: 30667592-30667592 on Build GRCh38
4643976 c.177C>T p.V59V Chr.16: 30667549-30667549 on Build GRCh38
3957474 c.2004C>T p.H668H Chr.16: 30667552-30667552 on Build GRCh38
1721221 c.2055G>A p.L685L Chr.16: 30667603-30667603 on Build GRCh38
6002197 c.2013C>G p.A671A Chr.16: 30667561-30667561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971482438 Chr.16: 30667592-30667592 on Build GRCh38
rs973577210 Chr.16: 30667713-30667713 on Build GRCh38
rs539429225 Chr.16: 30667552-30667552 on Build GRCh38
rs1000037008 Chr.16: 30667556-30667556 on Build GRCh38
rs756901081 Chr.16: 30667605-30667605 on Build GRCh38
rs1041929912 Chr.16: 30667901-30667901 on Build GRCh38
rs950829621 Chr.16: 30667719-30667719 on Build GRCh38
rs774653148 Chr.16: 30667575-30667575 on Build GRCh38
rs971541072 Chr.16: 30667732-30667732 on Build GRCh38
rs1054637332 Chr.16: 30667889-30667889 on Build GRCh38
rs545386226 Chr.16: 30667838-30667838 on Build GRCh38
rs531456749 Chr.16: 30667877-30667877 on Build GRCh38
rs1027695069 Chr.16: 30667714-30667714 on Build GRCh38
rs774077258 Chr.16: 30667514-30667514 on Build GRCh38
rs1023999980 Chr.16: 30667583-30667583 on Build GRCh38
rs1039377165 Chr.16: 30667486-30667486 on Build GRCh38
rs986645042 Chr.16: 30667866-30667866 on Build GRCh38
rs921878572 Chr.16: 30667802-30667802 on Build GRCh38
rs900314629 Chr.16: 30667495-30667495 on Build GRCh38
rs377542005 Chr.16: 30667846-30667846 on Build GRCh38
rs202186794 Chr.16: 30667591-30667591 on Build GRCh38
rs944361668 Chr.16: 30667578-30667578 on Build GRCh38
rs190690628 Chr.16: 30667740-30667740 on Build GRCh38
rs932212762 Chr.16: 30667871-30667871 on Build GRCh38
rs983843980 Chr.16: 30667758-30667758 on Build GRCh38
rs35795712 Chr.16: 30667741-30667741 on Build GRCh38
rs755398536 Chr.16: 30667662-30667662 on Build GRCh38
rs376987425 Chr.16: 30667571-30667571 on Build GRCh38
rs1020796036 Chr.16: 30667684-30667684 on Build GRCh38
rs559972712 Chr.16: 30667769-30667769 on Build GRCh38
rs891590521 Chr.16: 30667572-30667572 on Build GRCh38
rs945784652 Chr.16: 30667864-30667864 on Build GRCh38
rs1054312976 Chr.16: 30667847-30667847 on Build GRCh38
rs374123540 Chr.16: 30667553-30667553 on Build GRCh38
rs996412054 Chr.16: 30667620-30667620 on Build GRCh38
rs558888862 Chr.16: 30667891-30667892 on Build GRCh38
rs763535633 Chr.16: 30667580-30667580 on Build GRCh38
rs544074808 Chr.16: 30667680-30667680 on Build GRCh38
rs56133903 Chr.16: 30667857-30667857 on Build GRCh38
rs750037492 Chr.16: 30667641-30667641 on Build GRCh38
rs1010299815 Chr.16: 30667582-30667582 on Build GRCh38
rs959264850 Chr.16: 30667763-30667763 on Build GRCh38
rs778612975 Chr.16: 30667634-30667634 on Build GRCh38
rs183213523 Chr.16: 30667796-30667796 on Build GRCh38
rs1054524393 Chr.16: 30667637-30667637 on Build GRCh38
rs3747482 Chr.16: 30667638-30667638 on Build GRCh38
rs762127111 Chr.16: 30667643-30667643 on Build GRCh38
rs753368876 Chr.16: 30667602-30667602 on Build GRCh38
rs1033894121 Chr.16: 30667608-30667608 on Build GRCh38
rs566035880 Chr.16: 30667547-30667547 on Build GRCh38
rs867421871 Chr.16: 30667748-30667748 on Build GRCh38
rs541872105 Chr.16: 30667762-30667762 on Build GRCh38
rs983335519 Chr.16: 30667830-30667830 on Build GRCh38
rs946199039 Chr.16: 30667895-30667895 on Build GRCh38
rs574455309 Chr.16: 30667745-30667745 on Build GRCh38
rs907265404 Chr.16: 30667921-30667921 on Build GRCh38

View Full Product Details